Metaphosphoric Acid
m-Phosphoric Acid (Cryst. or Flakes/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: m-Phosphoric Acid CAS: 37267-86-0, 50813-16-6 CAS: 50813-16-6 Purity Grade: Certified ACS Physical Form: Solid Assay: 33.5 to 36.5% Assay Percent Range: 33.5 to 36.5% Molecular Formula: HPO3 MDL Number: MFCD00011351 Synonym: Glacial Phosphoric Acid
Metaphosphoric acid, Acros Organics
Chemical Name or Material: meta-Phosphoric Acid CAS: 37267-86-0, 50813-16-6 CAS: 50813-16-6 Purity Grade: ACS Reagent Molecular Formula: HO3P Synonym: Phosphoric Acid, meta
Metaphosphoric acid, stabilized, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Metaphosphoric acid CAS: 37267-86-0 CAS: 50813-16-6 Assay: 40 to 46% (HPO3) Molecular Formula: HO3P Linear Formula: (HPO3)n Formula Weight: 79.98g/mol MDL Number: MFCD00011351 Synonym: Phosphoric acid, meta
  spinner