p-Toluenesulfonic Acid
1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole, 98%, ACROS Organics™
Chemical Name or Material: 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole CAS: 77451-51-5 CAS Min %: 97.5 CAS Max %: 100.0 Assay Percent Range: 98% Molecular Formula: C9H8N4O4S Formula Weight: 268.25
p-Toluenesulfonic acid monohydrate, 99%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Toluenesulfonic acid monohydrate Name Note: >98.5% (total TSA) CAS: 6192-52-5 Purity Grade: Extra Pure Assay Percent Range: 99% Molecular Formula: C7H8O3S·H2O Linear Formula: CH3C6H4SO3H·H2O Formula Weight: 190.22 MDL Number: MFCD00142137 Synonym: PTSA
p-Toluenesulfonic Acid Monohydrate (Crystalline/Certified) ACS, Fisher Chemical
Chemical Name or Material: p-Toluenesulfonic Acid Monohydrate CAS: 6192-52-5 Purity Grade: Certified Grade Physical Form: Solid Molecular Formula: C7H8O3S · H2O Linear Formula: CH3C6H4SO3H·H2O Formula Weight: 190.22 MDL Number: MFCD00142137
p-Toluenesulfonic acid monohydrate, ACS reagent, ACROS Organics™
Chemical Name or Material: p-Toluenesulfonic acid monohydrate CAS: 6192-52-5 Purity Grade: ACS Reagent Grade Assay: ≥98.5% (Acid) Assay Percent Range: 99% Molecular Formula: C7H8O3S·H2O Linear Formula: CH3C6H4SO3H·H2O Formula Weight: 190.22 MDL Number: MFCD00142137 Synonym: PTSA
p-Toluenesulfonic acid monohydrate, ≥99%, MP Biomedicals™
Chemical Name or Material: p-Toluenesulfonic acid monohydrate CAS: 190.21 Physical Form: Powder Assay Percent Range: ≥99% Formula Weight: 190.21 Synonym: Toluene sulfonic acid
p-Toluenesulfonic Acid, Monohydrate, BAKER™, J.T.Baker™
Chemical Name or Material: p-Toluenesulfonic Acid CAS: 6192-52-5 Name Note: Monohydrate Molecular Formula: CH3C6H4SO3H·H2O Formula Weight: 190.22
p-Toluenesulfonic Acid, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: p-Toluenesulfonic Acid CAS: 06192-52-5 Name Note: Monohydrate Purity Grade: BAKER ANALYZED™ Reagent Molecular Formula: CH3C6H4SO3H·H2O Formula Weight: 190.22
p-Toluenesulfonic acid, Monohydrate, 99.0% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: p-Toluenesulfonic acid, Monohydrate Name Note: For Synthesis CAS: 6192-52-5 Physical Form: Hygroscopic Crystals Assay Percent Range: 99.0% min. Molecular Formula: C7H8O3S · H2O Linear Formula: CH3C6H4SO3H · H2O Formula Weight: 19.22 Synonym: PTSA
  spinner