Pamoic Acid
Pamoic acid disodium salt, Tocris™
Chemical Name or Material: Pamoic acid disodium salt CAS: 6640-22-8 Assay Percent Range: >99% Molecular Formula: C23H14Na2O6 Formula Weight: 432.33g/mol Synonym: 4, 4'-Methylenebis(3-hydroxy-2-naphthalenecarboxylic acid) disodium salt
Pamoic acid, 98+%, ACROS Organics™
Chemical Name or Material: Pamoic acid CAS: 130-85-8 Assay: 98.0% min. Assay Percent Range: 98+% Molecular Formula: C23H16O6 Formula Weight: 388.38g/mol MDL Number: MFCD00004079 Synonym: 4, 4';-Methylenebis[3-hydroxy-2-naphthalenecarboxylic acid]
Pamoic acid, 99%
Chemical Name or Material: Pamoic Acid CAS: 130-85-8 Molecular Formula: C23H16O6 Formula Weight: 388.38g/mol MDL Number: MFCD00004079 Beilstein: 901319 Merck Index: 14,7005
  spinner