Perchloric Acid
Perchloric Acid, 60% (Certified ACS), Fisher Chemical
Chemical Name or Material: Perchloric Acid, 60% CAS: 7601-90-3, 7732-18-5 CAS: 7732-18-5 Purity Grade: Certified ACS Assay: 60.0 to 62.0 w/w% Assay Percent Range: 60.0 to 62.0 w/w % Molecular Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric Acid, 70% (Certified ACS), Fisher Chemical
Chemical Name or Material: Perchloric Acid, 70% CAS: 7601-90-3 CAS: 7732-18-5 Purity Grade: Certified ACS Assay: 69.0 to 72.0 w/w% Assay Percent Range: 69.0 to 72.0 w/w % Molecular Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric Acid Solution, N/10 (0.1N) (Certified), Fisher Chemical
Chemical Name or Material: Perchloric Acid Solution Name Note: N/10 (0.1N) CAS: 64-19-7 CAS: 7601-90-3 Purity Grade: Certified Molecular Formula: ClHO4 MDL Number: MFCD00011325
Perchloric Acid, 0.02 Normal in Glacial Acetic Acid, Ricca Chemical
Chemical Name or Material: Perchloric Acid CAS: 64-19-7 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 108-24-7 CAS Min %: 2.07 CAS Max %: 2.15 CAS: 7601-90-3 CAS Min %: 0.26 CAS Max %: 0.27 Physical Form: Clear, colorless to light-yellow liquid
Perchloric Acid, 0.0011 Normal in Glacial Acetic Acid, Ricca Chemical
Chemical Name or Material: Perchloric Acid CAS: 64-19-7 CAS Min %: 96.0 CAS Max %: 100.0 CAS: 108-24-7 CAS Min %: 2.07 CAS Max %: 2.16 CAS: 7601-90-3 CAS Min %: 0.01 CAS Max %: 0.01 Physical Form: Clear, colorless to light-yellow liquid
Perchloric Acid, 0.10N in Glacial Acetic Acid) Volumetric Solution, Standard, Macron Fine Chemicals™
Chemical Name or Material: Perchloric Acid, 0.10 Normal (in Glacial Acetic Acid) Volumetric Solution CAS: 7601-90-3 Physical Form: Liquid
Perchloric acid, Acros Organics
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3, 7732-18-5 CAS: 7732-18-5 Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric Acid, 0.7, ULTREX™ II Ultrapure Reagent, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Purity Grade: ULTREX™ II Ultrapure Reagent Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric Acid, 0.7, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Purity Grade: BAKER INSTRA-ANALYZED™ Plus Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric Acid, Normality about 12, 69 to 72%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Name Note: Normality about 12 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric acid, for analysis, ca. 70% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Perchloric acid Name Note: p.a. CAS: 7601-90-3 CAS: 7732-18-5 Purity Grade: For analysis Assay: 70 to 72% Molecular Formula: HClO4 Linear Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric Acid, 8% w/v Aqueous, Ricca Chemical
Chemical Name or Material: Perchloric Acid CAS: 7732-18-5 CAS Min %: 88.0 CAS Max %: 92.0 CAS: 7601-90-3 CAS Min %: 9.97 CAS Max %: 10.38 Physical Form: Clear colorless liquid
Perchloric Acid, 0.1N in Glacial Acetic Acid Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Name Note: 0.1N in Glacial Acetic Acid Volumetric Solution Purity Grade: BAKER ANALYZED™ Reagent Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric Acid, 69 to 72%, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Purity Grade: BAKER INSTRA-ANALYZED™ Reagent Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric Acid, ca. 70% Solution in Water, For analysis, ACROS Organics™
Chemical Name or Material: Perchloric acid Name Note: ca. 70% Solution in Water CAS: 7601-90-3 CAS: 7732-18-5 Purity Grade: ACS Reagent Assay: 69.0 to 72.0% Molecular Formula: HClO4 Linear Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric acid, pure, 0.1M solution in acetic acid, ACROS Organics™
Chemical Name or Material: Perchloric acid Name Note: 0.1 M Solution in Acetic Acid CAS: 7601-90-3 CAS: 64-19-7 Purity Grade: Pure Molecular Formula: ClHO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric Acid, 2.00 N Aqueous, Ricca Chemical
Chemical Name or Material: Perchloric Acid CAS: 7732-18-5 CAS Min %: 76.0 CAS Max %: 79.0 CAS: 7601-90-3 CAS Min %: 21.71 CAS Max %: 22.59 Physical Form: Clear colorless liquid
Perchloric Acid, 60 to 62%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Perchloric acid, 0.1N in Acetic Acid Standardized Solution, Alfa Aesar™
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Physical Form: Liquid Molecular Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric acid, ACS, 70%, redistilled
Chemical Name or Material: Perchloric Acid, redistilled CAS: 7601-90-3 Name Note: Redistilled Purity Grade: ACS Molecular Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325 UN Number: UN1873 Merck Index: 14,7153
Perchloric acid, ACS, 48-50%, Alfa Aesar™
Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm
Perchloric acid, 70%, 99.9985% (metals basis), Alfa Aesar™
Chemical Name or Material: Perchloric Acid 70% CAS: 7601-90-3 Physical Form: Liquid Assay Percent Range: 99.9985% (metals basis) Molecular Formula: HClO4 Formula Weight: 100.46g/mol MDL Number: MFCD00011325
Perchloric Acid, ACS, 60-62%, Alfa Aesar
Chemical Name or Material: Perchloric Acid CAS: 7601-90-3 Molecular Formula: HClO4 Formula Weight: 100.46g/mol
Peracetic Acid, Quant™ Test Strips, Alfa Aesar™
Chemical Name or Material: Peracetic acid Name Note: 100 Strips to measure 5-50ppm peracetic acid
Perchloric Acid, in Glacial Acetic Acid, for Non-Aqueous Titrations, Certified, 0.1000N ±0.0005N, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Perchloric Acid CAS: 64-19-7 CAS Max %: 97.04 CAS: 108-24-7 CAS Max %: 2.1 CAS: 7601-90-3 CAS Max %: 0.86 Name Note: in Glacial Acetic Acid, for Non-Aqueous Titrations Purity Grade: Certified Linear Formula: HClO4 Formula Weight: 100.47g/mol UN Number: UN2789
  spinner