Phosphoric Acid
o-Phosphoric Acid, 85% (Certified ACS), Fisher Chemical
Chemical Name or Material: o-Phosphoric Acid Name Note: 85% CAS: 7664-38-2 CAS: 7732-18-5, 7732-18-5 Purity Grade: Certified ACS Assay: 85.0 w/w% min. Assay Percent Range: ≥85.0 w/w % Molecular Formula: H3O4P Formula Weight: 98g/mol MDL Number: MFCD00011340 Synonym: Orthophosphoric Acid
o-Phosphoric Acid, 85% (HPLC), Fisher Chemical
Chemical Name or Material: o-Phosphoric Acid Name Note: 85% CAS: 7664-38-2, 7732-18-5 CAS: 7732-18-5 Purity Grade: HPLC Purity Grade Notes: Also meets ACS specifications Assay: 85.0% min. Assay Percent Range: ≥85.0 % Molecular Formula: H3O4P Formula Weight: 98g/mol MDL Number: MFCD00011340
o-Phosphoric Acid (NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: o-Phosphoric Acid CAS: 7664-38-2, 7732-18-5 CAS: 7732-18-5 Purity Grade: NF/FCC Assay: 85.0 to 88.0% (NF) Molecular Formula: H3O4P Formula Weight: 98g/mol MDL Number: MFCD00011340
o-Phosphoric Acid (NF/FCC/EP/BP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: o-Phosphoric Acid CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: NF/FCC/EP/BP Assay: 84.0 to 90.0% (EP) Molecular Formula: H3O4P Formula Weight: 98g/mol MDL Number: MFCD00011340
Phosphoric acid, J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: H3PO4 Formula Weight: 97.994g/mol Synonym: Orthophosphoric Acid; Phosphoric(V) acid
Phosphoric acid, for analysis, 85 wt% solution in water, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Phosphoric acid Name Note: 85 wt% Solution in Water CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: For analysis Assay: 85% min. Molecular Formula: H3O4P Linear Formula: H3PO4 Formula Weight: 98.00g/mol MDL Number: MFCD00011340 Synonym: Orthophosphoric acid
Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid NF/FCC, Macron Fine Chemicals™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: NF/FCC Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid, N.F. - F.C.C., J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: NF, FCC Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid, 50% v/v, Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 7664-38-2 CAS Min %: 49.0 CAS Max %: 51.0 Physical Form: Clear colorless liquid
Phosphoric acid, pure, ACROS Organics™
Chemical Name or Material: Phosphoric acid CAS: 7664-38-2 Purity Grade: Pure Assay: 98.0% min. Molecular Formula: H3O4P Linear Formula: H3PO4 Formula Weight: 98.00g/mol MDL Number: MFCD00011340 Synonym: Orthophosphoric acid
meta-Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: meta-Phosphoric Acid CAS: 37267-86-0 Purity Grade: BAKER ANALYZED™ ACS Reagent Purity Grade Notes: Meets ACS reagent chemical requirements
Phosphoric Acid, 20% (v/v) Aqueous Solution (1 + 4), Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7732-18-5 CAS Min %: 79.0 CAS Max %: 81.0 CAS: 7664-38-2 CAS Min %: 19.0 CAS Max %: 21.0 Physical Form: Clear colorless liquid
Phosphoric Acid, ULTREX™ II Ultrapure Reagent, J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: ULTREX™ II Ultrapure Reagent Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric acid, J.T.Baker™
Chemical Name or Material: Phosphoric acid CAS: 7664-38-2 Physical Form: Solid or Viscous Liquid Molecular Formula: H3PO4 Formula Weight: 97.994g/mol Synonym: Orthophosphoric acid; Phosphoric(V) acid
Phosphoric acid, extra pure, 85% solution in water, ACROS Organics™
Chemical Name or Material: Phosphoric acid Name Note: 85% Solution in Water CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: Extra pure Assay: 84.5 to 86.0% Molecular Formula: H3O4P Linear Formula: H3PO4 Formula Weight: 98.00g/mol MDL Number: MFCD00011340 Synonym: Orthophosphoric acid
Phosphoric Acid, 10% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 91.0 CAS: 7664-38-2 CAS Min %: 9.0 CAS Max %: 11.0 Physical Form: Clear colorless liquid
Phosphoric Acid 85% Acid, ACS Reagent Grade, Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 CAS Min %: 85.0 CAS Max %: 100.0 CAS: 7732-18-5 (Water, Deionized) Purity Grade: ACS Reagent Physical Form: Clear colorless liquid
Phosphoric Acid, Diluted, 10% Solution, Multi-Compendial, N.F., J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Name Note: Diluted, 10% Solution Purity Grade: Multi-Compendial, NF Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid, 25% (v/v) Aqueous Solution (1 + 3), Ricca Chemical
Chemical Name or Material: Phosphoric Acid Name Note: 25% (v/v) CAS: 7664-38-2 CAS: 7732-18-5 Physical Form: Clear colorless liquid Assay Percent Range: 25.00 ± 0.04% (v/v)
Phosphoric Acid, Multi-Compendial, NF, J.T.Baker™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: Multi-Compendial, NF Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 Purity Grade: ACS Assay Percent Range: 85% Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric acid, ACS reagent, 85+% solution in water, ACROS Organics™
Chemical Name or Material: Phosphoric acid Name Note: 85+% Solution in Water CAS: 7664-38-2 CAS: 7732-18-5 Purity Grade: ACS Reagent Assay: 85.0% min. Molecular Formula: H3O4P Linear Formula: H3PO4 Formula Weight: 98.00g/mol MDL Number: MFCD00011340 Synonym: Orthophosphoric acid
Phosphoric Acid, 75% (v/v), Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 CAS: 7732-18-5
Phosphoric Acid, 85%, AR Grade, For ACS analysis, Macron Fine Chemicals™
CAS: 7664-38-2 Purity Grade: AR Purity Grade Notes: For ACS analysis (by acidimetry) Physical Form: Liquid Assay Percent Range: min. 85.0% Molecular Formula: H3PO4 Formula Weight: 98.00g/mol
Phosphoric Acid, 0.1% (v/v), Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7664-38-2 CAS: 7732-18-5
Phosphoric Acid, 10% (v/v) Aqueous Solution, Ricca
Chemical Name or Material: Phosphoric Acid
Phosphoric Acid, 50% v/v (1+1), Ricca Chemical
Chemical Name or Material: Phosphoric Acid Name Note: 50% v/v (1+1) CAS: 7732-18-5 CAS: 7664-38-2 Assay Percent Range: 50.0 ± 0.5% v/v
Phosphoric Acid Solution, 33% (w/w), Biotech Reagent, J.T.Baker™
Chemical Name or Material: Phosphoric Acid Solution CAS: 7664-38-2 Purity Grade: Biotech Reagent Molecular Formula: H3PO4 Formula Weight: 98g/mol
Phosphoric Acid, 30% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Phosphoric Acid CAS: 7732-18-5 CAS Min %: 69.0 CAS Max %: 71.0 CAS: 7664-38-2 CAS Min %: 29.0 CAS Max %: 31.0 Physical Form: Clear colorless liquid
  spinner