Phosphorous Acid
Phosphorous acid, 98%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Phosphorous acid CAS: 13598-36-2 Purity Grade: Extra Pure Assay: ≥97.5% Assay Percent Range: 98% Molecular Formula: H3O3P Linear Formula: H3PO3 Formula Weight: 82 MDL Number: MFCD00137258 Synonym: Orthophosphorous acid
Phosphorous acid, 97%
Chemical Name or Material: Phosphorous acid CAS: 13598-36-2 Molecular Formula: H3PO3 Formula Weight: 82 MDL Number: MFCD00137258 UN Number: UN2834 Merck Index: 14,7346 Synonym: Phosphonic acid
Phosphorous Acid, Honeywell Fluka™
CAS: 13598-36-2 Chemical Name or Material: Phosphorous Acid Density: 1.651g/mL at 25°C Assay: 99% Formula Weight: 82g/mol EINECS Number: 237-066-7 Health Hazard 2: P280-P305 + P351 + P338-P310 Health Hazard 3: H290-H302-H314 MDL Number: MFCD00137258 Molecular Formula: H3O3P RTECS Number: SZ6400000 UN Number: 2834
Phosphoric Acid, Certified, 10.0% (v/v) ±0.2% (v/v), LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Phosphoric Acid, (1+9) CAS: 7732-18-5 CAS Max %: 90 CAS: 7664-38-2 CAS Max %: 10 Purity Grade: Certified Grade Linear Formula: H3PO4 Formula Weight: 97.1 UN Number: UN1805
  spinner