Phthalic Acid
Phthalic acid, 99%, ACROS Organics™
Chemical Name or Material: Phthalic acid CAS: 88-99-3 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C8H6O4 Formula Weight: 166.13g/mol MDL Number: MFCD00002467 Synonym: 1, 2-Benzenedicarboxylic acid
Phthalic acid, ACS, 99.5+%
Chemical Name or Material: Phthalic acid CAS: 88-99-3 Purity Grade: For ACS analysis Molecular Formula: C6H4(COOH)2 Formula Weight: 166.13g/mol MDL Number: MFCD00002467 Beilstein: 608199 Merck Index: 14,7371 Synonym: Benzene-1, 2-dicarboxylic acid
Phthalic acid, 100.6%, For ACS analysis, MP Biomedicals™
Chemical Name or Material: Phthalic acid Name Note: For ACS analysis CAS: 88-99-3 Purity Grade: 1.006 Physical Form: Colorless Powder Assay Percent Range: ≥99.5% Molecular Formula: C8H6O4 Formula Weight: 166.1g/mol
  spinner