Picric Acid
Picric Acid, Saturated (AOAC), Fisher Chemical
Chemical Name or Material: Picric Acid, Saturated CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31
Picric Acid, Saturated Aqueous Solution, approximately 1.2% (w/v), Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31 Physical Form: Clear, yellow liquid
Trinitrophenol TS (Picric Acid TS), 1% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trinitrophenol TS (Picric Acid TS) CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 88-89-1 CAS Min %: 1.26 CAS Max %: 1.31 Physical Form: Clear, yellow liquid
Picric Acid, 4% (w/v) in Denatured Ethanol (Reagent Alcohol), Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 64-17-5 CAS Min %: 84.0 CAS Max %: 88.0 CAS: 88-89-1 CAS Min %: 4.67 CAS Max %: 4.86 CAS: 67-63-0 CAS Min %: 4.46 CAS Max %: 4.64 CAS: 88-89-1 (Picric Acid (2, 4, 6-Trinitrophenol)) Physical Form: Clear, yellow liquid
Picric Acid, Reagent Grade, Ricca Chemical
Chemical Name or Material: Picric Acid CAS: 88-89-1 CAS Min %: 98.0 CAS Max %: 100.0 CAS: 7732-18-5 CAS Min %: 29.0 CAS Max %: 31.0 Purity Grade: Reagent Physical Form: Yellow crystalline solid packed in 30% water
Picric Acid, Saturated, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Picric Acid CAS: 7732-18-5 CAS Max %: 98.7 CAS: 88-89-1 CAS Max %: 1.3 Name Note: Saturated Purity Grade: Certified Linear Formula: C6H3N3O7 Formula Weight: 229.1g/mol
  spinner