Sulfamic Acid
Sulfamic Acid, BAKER™, J.T.Baker™
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Purity Grade: BAKER™ Molecular Formula: NH2SO3H Formula Weight: 97.09g/mol
Sulfamic Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: NH2SO3H Formula Weight: 97.09g/mol
Sulfamic Acid, Practical grade, 97% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Purity Grade: Practical Physical Form: Crystalline Assay Percent Range: 97%, min. Molecular Formula: NH2SO3H Linear Formula: NH2SO3H Formula Weight: 97.9g/mol Synonym: Amidosulfonic Acid
Sulfamic Acid, 99.3 to 100.3%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 Name Note: For Synthesis Molecular Formula: H3NO3S Linear Formula: HCH2NSOCH3H Formula Weight: 97.8g/mol Synonym: Amidosulfonic acid
Sulfamic Acid, 99.3 to 100.3%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid Name Note: For Synthesis CAS: 5329-14-6 Physical Form: White crystals Assay Percent Range: 99.3 to 100.3% Molecular Formula: H3NO3S Linear Formula: HCH2NSOCH3H Formula Weight: 97.8g/mol Synonym: Amidosulfonic Acid
Sulfamic Acid, ACS Grade, 99.3 to 100.3%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sulfamic Acid CAS: 5329-14-6 CAS Max %: 100 Purity Grade: ACS Grade Linear Formula: NH2SO3H Formula Weight: 97.09g/mol UN Number: UN2967
  spinner