Sulfurous Acid
Sulfurous Acid (Certified ACS), Fisher Chemical
Chemical Name or Material: Sulfurous Acid CAS: 7782-99-2 Purity Grade: Certified ACS Grade Assay: ≥6.0% Molecular Formula: H2SO3 MDL Number: MFCD00011335 Synonym: Sulfur Dioxide Solution
Sulfurous Acid, ACS, 6.0% SO2 min, Alfa Aesar
Chemical Name or Material: Sulfurous Acid Name Note: 6.0% SO2 min. CAS: 7782-99-2 Purity Grade: ACS Grade Physical Form: Liquid Molecular Formula: H2SO3 Formula Weight: 82.08 MDL Number: MFCD00011335 Synonym: Sulfur Dioxide Solution
Sulfurous Acid ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Sulfurous Acid CAS: 7782-99-2 Purity Grade: ACS Grade
  spinner