Tartaric Acid
L-(+)-Tartaric Acid (Granular/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.0% min. Assay Percent Range: ≥99.0 % Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09g/mol MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
L-(+)-Tartaric Acid (Powder/Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.0% min. Assay Percent Range: ≥99.0 % Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09g/mol MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
L-(+)-Tartaric acid dipotassium salt, 97%, Acros Organics
Chemical Name or Material: L-(+)-Tartaric acid dipotassium salt Assay Percent Range: 97% Molecular Formula: C4H4K2O6 Formula Weight: 226.27g/mol
L-(+)-Tartaric Acid (Fine Granular/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: NF/FCC Physical Form: Solid Assay: 99.7 to 100.5% (NF) Molecular Formula: C4H6O6 Linear Formula: HOOC(CHOH)2COOH Formula Weight: 150.09g/mol MDL Number: MFCD00064207 Synonym: 2, 3-Dihydroxybutanedioic Acid
L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L+-Tartaric acid, disodium salt, dihydrate CAS: 6106-24-7 Purity Grade: ACS Reagent Assay: 99.0 to 101.0% Assay Percent Range: 99+% Molecular Formula: C4H4Na2O6·2H2O Linear Formula: (CHOHCOONa)2·2H2O Formula Weight: 230.08g/mol MDL Number: MFCD00150035 Synonym: Disodium tartrate
L(+)-Tartaric acid diammonium salt, 99%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L(+)-Tartaric acid diammonium salt Name Note: 99% CAS: 3164-29-2 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C4H12N2O6 Linear Formula: [-CH(OH)CO2NH4]2 Formula Weight: 184.15g/mol MDL Number: MFCD00013073 Synonym: Ammonium L-tartrate
Tartaric Acid, 50% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Tartaric Acid Name Note: Aqueous Solution CAS: 7732-18-5 CAS: 87-69-4 Physical Form: Clear, colorless to slightly yellow liquid Assay Percent Range: 50% (w/v)
Tartaric Acid ACS, Ricca Chemical
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 CAS Min %: 98.0 CAS Max %: 100.0 Purity Grade: ACS
Tartaric Acid, 2% (w/v) Aqueous Solution, Certified APHA Reagent, Ricca Chemical
Chemical Name or Material: Tartaric Acid Name Note: 2% (w/v) Aqueous Solution CAS: 7732-18-5 CAS: 87-69-4 Physical Form: Clear, colorless to slightly yellow liquid
Tartaric Acid NF/FCC Fine Granular, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: NF, FCC Physical Form: Fine Granular Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
Tartaric Acid ACS AR Powder, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: ACS Grade Physical Form: Powder Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
L-(+)-Tartaric Acid, Granular, N.F. - F.C.C., J.T.Baker™
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: NF, FCC Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
L(+)-Tartaric acid, 99+%, Acros Organics Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: L(+)-Tartaric acid Assay: 99.0% min. Assay Percent Range: 99+% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09g/mol Synonym: L(+)-Dihydroxysuccinic acid
DL-Tartaric acid, 99.5%, ACROS Organics™
Chemical Name or Material: DL-Tartaric acid Assay: 99.4% min. Assay Percent Range: 99.50% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09g/mol
L-(+)-Tartaric Acid, Granular, Multi-Compendial, N.F., J.T.Baker™
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: Multi-Compendial, NF Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
D(-)-Tartaric Acid (unnatural Tartaric Acid) 99%, ACROS Organics™
Chemical Name or Material: D(-)-Tartaric acid Name Note: 99% CAS: 147-71-7 Assay: 98.5% min. Assay Percent Range: 99% Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09g/mol MDL Number: MFCD00004238 Synonym: D(-)-Dihydroxysuccinic acid
d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: d-Tartaric Acid CAS: 87-69-4 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
L(+)-Tartaric acid, ACS reagent, Acros Organics
Chemical Name or Material: L(+)-Tartaric acid Name Note: 99+% CAS: 87-69-4 Purity Grade: ACS Reagent Assay: 99.0% min. Molecular Formula: C4H6O6 Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09g/mol MDL Number: MFCD00064207 Synonym: L(+)-Dihydroxysuccinic acid
Tartaric Acid Aqueous Solution, 50% (w/v), Ricca Chemical
CAS: 7732-18-5 Purity Grade: 50% Physical Form: Solution Assay: 50% (w/v) Assay Percent Range: 50% (w/v)
Tartaric Acid AR Granular, Macron Fine Chemicals™
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Physical Form: Granular Molecular Formula: HOCO(CHOH)2COOH Formula Weight: 150.09g/mol
L-(+)-Tartaric acid, ACS
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Purity Grade: ACS Grade Molecular Formula: HOOC(CHOH)2COOH Formula Weight: 150.09g/mol MDL Number: MFCD00064207 Beilstein: 1725147 Merck Index: 14,9070
L-(+)-Tartaric Acid, ≥97%, MP Biomedicals™
Chemical Name or Material: L-(+)-Tartaric Acid CAS: 87-69-4 Physical Form: Off-white Powder Assay Percent Range: ≥97% Molecular Formula: C4H6O6 Formula Weight: 150.1g/mol Synonym: L-2, 3-Dihydroxybutanedioic acid
Tartaric Acid, Certified, 10.0% (w/v) ±0.5% (w/v), LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tartaric Acid CAS: 7732-18-5 CAS Max %: 90.4 CAS: 87-69-4 CAS Max %: 9.6 Purity Grade: Certified Linear Formula: C4H6O6 Formula Weight: 150.09g/mol
Tartaric Acid, ACS Grade, ≥ 99.0%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 CAS Max %: 100 Purity Grade: ACS Grade Linear Formula: C4H6O6 Formula Weight: 150.09g/mol
L-(+)-Tartaric Acid, GR, ACS, EMD Millipore
Chemical Name or Material: L-(+)-Tartaric Acid Name Note: For ACS analysis CAS: 87-69-4 Physical Form: White solid Molecular Formula: C4H6O6 Formula Weight: 150.09g/mol Synonym: L-2, 3-Dihydroxybutanedioic acid
Tartaric Acid Products for Science Education
Chemical Name or Material: Tartaric Acid CAS: 87-69-4 Purity Grade: Laboratory Assay Percent Range: >99% w/v Linear Formula: HO2CCH(OH)CH(OH)CO2H Formula Weight: 150.09g/mol Synonym: Threaric Acid
  spinner