Thiodiglycolic Acid
Thiodiglycolic acid, 98%, ACROS Organics™
Chemical Name or Material: Thiodiglycolic acid Name Note: 98% CAS: 123-93-3 Assay: 97.5% min. Assay Percent Range: 98% Molecular Formula: C4H6O4S Linear Formula: S(CH2CO2H)2 Formula Weight: 150.15g/mol MDL Number: MFCD00004359 Synonym: Thiodiacetic acid
  spinner