Tiglic Acid
Tiglic Acid, 98%, MP Biomedicals™
Chemical Name or Material: Tiglic Acid CAS: 80-59-1 Assay Percent Range: 98% Molecular Formula: C5H8O2 Formula Weight: 100.1g/mol Synonym: trans-2, 3-Dimethylacrylic acid
  spinner