Trichloroacetic Acid
Trichloroacetic Acid (Crystalline/Certified ACS), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.0% min. Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39g/mol MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic Acid (Crystalline/Certified), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Certified ACS Physical Form: Solid Assay: 99.0% min. Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39g/mol MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic Acid, 10% w/v (AOAC), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Name Note: 10% (w/v) CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 76-03-9 CAS Min %: 8.9 CAS Max %: 9.26 CAS: 7440-44-0 CAS Min %: 0.18 CAS Max %: 0.19
Trichloroacetic Acid, 15% w/v (AOAC), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Name Note: 15% (w/v) CAS: 7732-18-5 CAS Min %: 85.0 CAS Max %: 89.0 CAS: 76-03-9 CAS Min %: 12.77 CAS Max %: 13.29 CAS: 7440-44-0 CAS Min %: 0.17 CAS Max %: 0.18
Trichloroacetic Acid Solution (20% w/v), Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid Solution Name Note: 20% (w/v) CAS: 76-03-9 CAS: 7732-18-5 Assay: 20 to 21% Assay Percent Range: 20 to 21 % Molecular Formula: Cl3CCOOH MDL Number: MFCD00004177
Trichloroacetic Acid, Fisher Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 CAS: 7732-18-5 Assay: 99.5 to 101.0% Assay Percent Range: 99.5 to 101.0 % Molecular Formula: Cl3CCOOH Formula Weight: 163.39g/mol MDL Number: MFCD00004177
Trichloroacetic Acid, 10% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 89.0 CAS Max %: 93.0 CAS: 76-03-9 CAS Min %: 8.9 CAS Max %: 9.26 CAS: 7440-44-0 CAS Min %: 0.18 CAS Max %: 0.19
Trichloroacetic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: BAKER ANALYZED™ ACS Reagent Molecular Formula: Cl3CCOOH Formula Weight: 163.39g/mol
Trichloroacetic Acid, 80% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 54.0 CAS Max %: 57.0 CAS: 76-03-9 CAS Min %: 43.52 CAS Max %: 45.3 CAS: 7440-44-0 CAS Min %: 0.11 CAS Max %: 0.11 Physical Form: Clear, colorless to tan liquid
Trichloroacetic acid, sodium salt, 97%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic acid, sodium salt Assay: 96.0% min. Assay Percent Range: 97% Molecular Formula: C2Cl3NaO2 Linear Formula: CCl3CO2Na Formula Weight: 185.37g/mol MDL Number: MFCD00064198 Synonym: Sodium trichloroacetate
Trichloroacetic acid, 15% (w/v) aqueous solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic acid CAS: 7732-18-5 CAS: 76-03-9
Trichloroacetic acid, 20% (w/v) aqueous solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic acid CAS: 7732-18-5 CAS: 76-03-9
Trichloroacetic acid, 99+%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic acid Name Note: 99+% CAS: 76-03-9 Purity Grade: ACS Reagent Assay: 99.0% min. Assay Percent Range: 99+% Molecular Formula: C2HCl3O2 Linear Formula: CCl3CO2H Formula Weight: 163.39g/mol MDL Number: MFCD00004177 Synonym: TCA
Trichloroacetic Acid ACS AR Crystal, Macron Fine Chemicals™
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: ACS Grade Physical Form: Crystalline Molecular Formula: Cl3CCOOH Formula Weight: 163.39g/mol
Trichloroacetic Acid, 30% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 75.0 CAS Max %: 78.0 CAS: 76-03-9 CAS Min %: 22.59 CAS Max %: 23.51 CAS: 7440-44-0 CAS Min %: 0.15 CAS Max %: 0.16 Physical Form: Clear colorless liquid
Trichloroacetic Acid (Flakes or Crystals), Fisher BioReagents
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Physical Form: Solid Assay: 99.0% min. Assay Percent Range: ≥99.0 % Molecular Formula: C2HCl3O2 Linear Formula: CCl3COOH Formula Weight: 163.39g/mol Synonym: TCA
Trichloroacetic Acid, 33% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 74.0 CAS Max %: 77.0 CAS: 76-03-9 CAS Min %: 24.29 CAS Max %: 25.28 CAS: 7440-44-0 CAS Min %: 0.15 CAS Max %: 0.15 Physical Form: Clear colorless liquid
Trichloroacetic Acid, 3% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid
Trichloroacetic Acid, 5% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid
Trichloroacetic Acid, 24% (w/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Min %: 79.0 CAS Max %: 82.0 CAS: 76-03-9 CAS Min %: 18.95 CAS Max %: 19.72 CAS: 7440-44-0 CAS Min %: 0.16 CAS Max %: 0.16 Physical Form: Clear colorless liquid
Trichloroacetic Acid, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid Name Note: For ACS analysis CAS: 76-03-9 Physical Form: Colorless, deliquescent crystals Assay Percent Range: 99.5% min. Molecular Formula: CCl3COOH Formula Weight: 163.39g/mol
Trichloroacetic acid, 99.6%, MP Biomedicals™
Chemical Name or Material: Trichloroacetic acid CAS: 76-03-9 Physical Form: White crystalline powder Assay Percent Range: 0.996 Molecular Formula: C2HCl3O2 Formula Weight: 163.4g/mol Synonym: TCA
Trichloroacetic Acid Solution (6.1N (100% w/v))
Chemical Name or Material: Trichloroacetic Acid Solution Name Note: 6.1N CAS: 76-03-9 Physical Form: 100% w/v Formula Weight: 163.38g/mol
Trichloroacetic acid, ACS, ≥99%, MP Biomedicals™
Chemical Name or Material: Trichloroacetic acid CAS: 76-03-9 Physical Form: White crystalline powder Assay Percent Range: ≥99% Molecular Formula: C2HCl3O2 Formula Weight: 163.4g/mol Synonym: TCA
Trichloroacetic Acid, Certified, 10.0% (w/v) ±0.5%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Max %: 90 CAS: 76-03-9 CAS Max %: 10 Purity Grade: Certified Linear Formula: C2HCl3O2 Formula Weight: 163.39g/mol UN Number: UN2564 Synonym: TCA
Trichloroacetic Acid, Certified, 80.0% (w/v) ±2.0%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 CAS Max %: 80 CAS: 7732-18-5 CAS Max %: 20 Purity Grade: Certified Linear Formula: C2HCl3O2 Formula Weight: 163.39g/mol UN Number: UN2564 Synonym: TCA
Trichloroacetic Acid, Certified, 5.0% (w/v) ±0.4%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Max %: 95 CAS: 76-03-9 CAS Max %: 5 Purity Grade: Certified Linear Formula: C2HCl3O2 Formula Weight: 163.39g/mol UN Number: UN2564 Synonym: TCA
Trichloroacetic Acid, Certified, 3.0% (w/v) ±0.3%, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 7732-18-5 CAS Max %: 97 CAS: 76-03-9 CAS Max %: 3 Purity Grade: Certified Linear Formula: C2HCl3O2 Formula Weight: 163.39g/mol UN Number: UN2564 Synonym: TCA
Trichloroacetic Acid ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: ACS Linear Formula: CCl3COOH Formula Weight: 163.39g/mol
Trichloroacetic Acid Products for Science Education
Chemical Name or Material: Trichloroacetic Acid CAS: 76-03-9 Purity Grade: Laboratory Assay Percent Range: >95% Linear Formula: CCl3COOH Formula Weight: 163.39g/mol Synonym: Tca
  spinner