Trifluoroacetic Acid
Trifluoroacetic acid, 99.5%, for biochemistry, ACROS Organics™
Chemical Name or Material: Trifluoroacetic acid CAS: 76-05-1 Purity Grade: For biochemistry Assay: ≥99.5% Assay Percent Range: 99.5% Molecular Formula: C2HF3O2 Linear Formula: CF3CO2H Formula Weight: 114.02 MDL Number: MFCD00004169 Synonym: TFA
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Additional Information: Vapor Pressure: 107mbar at 25°C; Vapor Density: 3.9 Aluminum (Al): 20 ppb max. Assay Percent Range: ≥99.5 % Calcium (Ca): 50 ppb max. CAS: 76-05-1 ChemAlert Storage Symbol: White Chemical Name or Material: Trifluoroacetic Acid Chloride: Report ppm Copper (Cu): 10 ppb max. Density: 1.535 Assay: ≥99.5% Boiling Point: 72°C Color: Clear Colorless Formula Weight: 114.02 Melting Point: −15°C pH: 0.45 Physical Form: Liquid Purity Grade: Optima™ LC/MS DOT Information: DOT Class 8, : Corrosive Fluoride: Report ppm Health Hazard 2: DANGER! Health Hazard 3: Emergency Overview
Causes severe burns by all exposure routes. Inhalation may cause central nervous system effects. Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:0
Instability:0 Identification: Pass Test Iron (Fe): 10 ppb max. LCMS Gradient Suitability: Pass Test Lead (Pb): 10 ppb max. Linear Formula: CF3COOH Magnesium (Mg): 10 ppb max. Manganese (Mn): 10 ppb max. Merck Index: 15, 9847 MDL Number: 4169 Molecular Formula: C2HF3O2 Nickel (Ni): 10 ppb max. Potassium (K): 20 ppb max. Residue after Evaporation: 1ppm max. Silver (Ag): 10 ppb max. Sodium (Na): 50 ppb max. Sulfate: Report ppm Synonym: TFA Vapor Density: 3.9 Vapor Pressure: 107mbar 25 Water: 0.05% max. Zinc (Zn): 20 ppb max.
Trifluoroacetic acid, 99.5+%
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Molecular Formula: CF3CO2H Linear Formula: CF3CO2H Formula Weight: 114.02 MDL Number: MFCD00004169 UN Number: UN2699 Beilstein: 742035 Merck Index: 14,9681
Trifluoroacetic Acid, 99%, Alfa Aesar™
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: CF3CO2H Formula Weight: 114.02
Trifluoroacetic Acid (For Peptide and Protein Analysis/Certified) ACS, Fisher Chemical
Chemical Name or Material: Trifluoroacetic Acid Name Note: For Peptide and Protein Analysis CAS: 76-05-1 Purity Grade: Certified Grade Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C2HF3O2 Linear Formula: CF3COOH Formula Weight: 114.02 MDL Number: MFCD00004169
Trifluoroacetic Acid (Clear, Fuming Liq./Biotechnology Grade), Fisher Chemical
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Purity Grade: Biotech Grade Physical Form: Liquid Assay: ≥98.5% Assay Percent Range: ≥98.5 % Molecular Formula: C2HF3O2 Linear Formula: CF3COOH Formula Weight: 114.02 MDL Number: MFCD00004169
Trifluoroacetic Acid (Reagent), Fisher Chemical
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Purity Grade: Reagent Grade Assay: ≥97% Assay Percent Range: ≥97 % Molecular Formula: C2HF3O2 Linear Formula: CF3COOH Formula Weight: 114.02 MDL Number: MFCD00004169 Synonym: TFA
Trifluoroacetic Acid, BAKER ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Purity Grade: BAKER ANALYZED™ Reagent Molecular Formula: F3CCOOH Formula Weight: 114.03
Trifluoroacetic Acid, HPLC Grade, J.T.Baker™
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Name Note: Gradient Tested for HPLC Peptide and Protein Analysis Purity Grade: HPLC Grade Molecular Formula: F3CCOOH Formula Weight: 114.03
Trifluoroacetic Acid (Peptide Synthesis Grade), Fisher BioReagents™
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Purity Grade: Peptide Synthesis Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C2HF3O2 Linear Formula: CF3COOH Formula Weight: 114.02 Synonym: Perfluoroacetic Acid
Trifluoroacetic acid, ReagentPlus™, Honeywell
Beilstein: 742035 CAS: 76-05-1 Chemical Name or Material: Trifluoroacetic acid Density: 1.489g/mL at 20°C Assay: 99% Formula Weight: 114.02g/mol Purity Grade: ReagentPlus™ Flash Point: >100°C (>212°F) Health Hazard 2: P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 Linear Formula: CF3COOH MDL Number: MFCD00004169 RTECS Number: AJ9625000 UN Number: UN2699 Vapor Density: 3.9 (vs air) Vapor Pressure: 97.5 mmHg (20°C)
Trifluoroacetic Acid, HPLC, 99.9% min., EMD Millipore
Chemical Name or Material: Trifluoroacetic Acid Name Note: For Protein Sequence Analysis CAS: 76-05-1 Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: C2HF3O2 Linear Formula: CF3COOH Formula Weight: 114.2 Synonym: TFA
Trifluoroacetic Acid, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Trifluoroacetic Acid CAS: 76-05-1 Purity Grade: NF/USP Physical Form: Liquid Linear Formula: CF3COOH Formula Weight: 114.2 Synonym: Perfluoroacetic Acid
Trifluoroacetic Acid, For protein sequencing, Honeywell Fluka™
Additional Information: For protein sequencing Beilstein: 742035 CAS: 76-05-1 Chemical Name or Material: Trifluoroacetic Acid Density: 1.489g/mL at 20°C Assay: ≥99% Formula Weight: 114.02g/mol EINECS Number: 200-929-3 Flash Point: >100°C (>212°F) Health Hazard 2: P273-P280-P305 + P351 + P338-P310 Health Hazard 3: H314-H332-H412 Linear Formula: CF3COOH MDL Number: MFCD00004169 RTECS Number: AJ9625000 UN Number: 2699 Vapor Density: 3.9 (vs air) Vapor Pressure: 97.5mmHg (20°C)
  spinner