Chromatography Media
Cytiva MabSelect Sure™ Chromatography Media <img src=
Cytiva MabSelect Sure™ Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
MabSelect™ SuRe™ is a protein A affinity medium for capturing MAbs at process scale with an alkali-tolerant rProtein A ligand that allows the for cost-effective sanitization protocols
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q <img src=
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q Available on GSA/VA Contract for Federal Government customers only.
Use for preparative protein separations in both research and industrial applications. Cytiva Q Sepharose™ Fast Flow Chromatography Media is a strong anion exchanger based on the well established Sepharose Fast Flow ion exchange platform.
Cytiva HiTrap™ TALON™ Crude Prepacked Columns <img src=
Cytiva HiTrap™ TALON™ Crude Prepacked Columns Available on GSA/VA Contract for Federal Government customers only.
For purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC).   Cytiva Lifescience™ HiTrap™ TALON™ Crude Prepacked Columns offer a cobalt-based medium allowing protein purification under native or denaturing conditions.
Cytiva Butyl Sepharose™ High Performance Chromatography Media
Cytiva Butyl Sepharose™ High Performance Chromatography Media
Designed for intermediate and polishing step purification steps
Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media <img src=
Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Used for purification of monoclonal and polyclonal antibodies.   Cytiva Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.
Cytiva Blue Sepharose™ 6 Fast Flow Chromatography Media
Cytiva Blue Sepharose™ 6 Fast Flow Chromatography Media
Blue dye binds to proteins such as albumin and lipoproteins as well as enzymes like kinases and dehydrogenases
Cytiva Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media <img src=
Cytiva Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
USe for  separations of biomolecules with an affinity for heparin, including antithrombin III, coagulation factors and other plasma proteins.  Cytiva Life Sciences™ Heparin Sepharose™ 6 Fast Flow Affinity Chromatography Media is fast and reliable and highly suitable for process-scale purification processes.
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: DEAE
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: DEAE
Used for preparative protein separations in both research and industrial applications.  Cytiva Life Sciences™ Sephaorose™ Fast Flow Anion Exchange Media: DEAE ensures low levels of non-specific binding.
Cytiva Q Sepharose™ High Performance Chromatography Media <img src=
Cytiva Q Sepharose™ High Performance Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Designed for high intermediate and polishing steps in downstream processing
Cytiva PlasmidSelect Xtra Medium <img src=
Cytiva PlasmidSelect Xtra Medium Available on GSA/VA Contract for Federal Government customers only.
PlasmidSelect Xtra is a thiophilic aromatic adsorption chromatography resin with a selectivity that allows supercoiled covalently closed circular forms of plasmid DNA to be separated from open circular forms.
Cytiva rProtein A Sepharose™ Fast Flow Affinity Media <img src=
Cytiva rProtein A Sepharose™ Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Designed for purification of monoclonal and polyclonal antibodies at labs and process scales
Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media <img src=
Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Use for coupling amino-containing smaller proteins and peptides.  Cytiva Life Sciences™ NHS-Activated Sepharose™ 4 Fast Flow Affinity Media yields coupled products that are very stable, especially at high pH.
Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media <img src=
Cytiva Phenyl Sepharose™ 6 Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Used for capture and intermediate purification requiring low to medium hydrophobicity
Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium
Cytiva Capto™ MMC ImpRes Multimodal BioProcess™ Chromatography Medium
For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules
Cytiva Phenyl Sepharose™ High Performance Media
Cytiva Phenyl Sepharose™ High Performance Media
Designed for intermediate and polishing step purification steps
Cytiva Sepharose™ Fast Flow Cation Exchange Media <img src=
Cytiva Sepharose™ Fast Flow Cation Exchange Media Available on GSA/VA Contract for Federal Government customers only.
SP Sepharose Fast Flow is a sulphopropyl (SP) strong cation exchange chromatography resin for fast protein purification. CM Sepharose Fast Flow is a weak cation exchanger based on the well established Sepharose Fast Flow ion exchange platform, extensively used for preparative protein separations in both research and industrial applications.
Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin
Cytiva Strep-Tactin™ XT Sepharose™ Chromatography Resin
Chromatography resin for purification of recombinant proteins tagged with a Strep-tag™ II and Twin-Strep-tag™.
Cytiva TALON™ Superflow Medium
Cytiva TALON™ Superflow Medium
Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.
Cytiva CNBr-Activated Sepharose™ 4 Fast Flow Affinity Media <img src=
Cytiva CNBr-Activated Sepharose™ 4 Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.
Used for coupling of large amino-containing ligands
Cytiva HiTrap™ AVB Sepharose™ Columns <img src=
Cytiva HiTrap™ AVB Sepharose™ Columns Available on GSA/VA Contract for Federal Government customers only.
Enables purification of adeno associated viruses (AAV) in one step, with high purity and excellent yields. Cytiva HiTrap™ AVB Sepharose™ Columns are pre-packed with affinity chromatography medium (resin) for simple operation with a syringe, pump or chromatographic system.
Cytiva DEAE Sepharose™ Fast Flow Chromatography Media <img src=
Cytiva DEAE Sepharose™ Fast Flow Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
For preparative protein separations in both research and industrial applications. Cytiva Life Sciences™ DEAE Sepharose™ Fast Flow Chromatography Media can be used for capture and intermediate purification of wide range of approved biopharmaceuticals.
Cytiva MabSelect™ Chromatography Media <img src=
Cytiva MabSelect™ Chromatography Media Available on GSA/VA Contract for Federal Government customers only.
Protein A affinity medium designed for capturing MAbs from large sample volumes
Cytiva SOURCE™ 30S Chromatography Media
Cytiva SOURCE™ 30S Chromatography Media
Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides
Cytiva Capto™ Q Chromatography Media
Cytiva Capto™ Q Chromatography Media
Designed for capture and intermediate purification of proteins from large feed volumes. Cytiva CaptoTM Q Chromatography Media is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers.
Cytiva SP Sepharose™ XL
Cytiva SP Sepharose™ XL
Strong cation exchanger with high loading capacity and chemical stability
Cytiva Capto™ Blue Chromatography Media
Cytiva Capto™ Blue Chromatography Media
An affinity medium for capture of proteins and enzyme purification on large and small scale
Cytiva Capto™ Adhere Media
Cytiva Capto™ Adhere Media
A multimodal anion exchanger to follow the Protein A capture step, making for a streamlined process
Cytiva Capto™ Multimodal (MMC) Chromatography Media
Cytiva Capto™ Multimodal (MMC) Chromatography Media
Used to solve specific purification challenges at both high and low ionic strengths
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: Q
Use for preparative protein separations in both research and industrial applications. Cytiva Q Sepharose™ Fast Flow Chromatography Media is a strong anion exchanger based on the well established Sepharose Fast Flow ion exchange platform.
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: ANX
Cytiva Sephaorose™ Fast Flow Anion Exchange Media: ANX
Used for preparative protein separations in both research and industrial applications
  spinner