Derivation and Visualization Reagents
Thermo Scientific™ Pierce™ Triethylamine (TEA), Sequencing Grade
Bottles of 99.5%-pure TEA (MW 101.2, CAS 121-44-8) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
  spinner