HPLC Ion Pair Reagents
Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade
Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
Thermo Scientific™ Pierce™ Formic Acid, LC-MS Grade Available on GSA/VA Contract for Federal Government customers only.
A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.
Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), LC-MS Grade
Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), Sequencing Grade
Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), HPLC Grade
Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).
  spinner