Karl Fischer Reagents
CombiCoulomat Fritless Reagent, Apura™, EMD Chemicals Available on GSA/VA Contract for Federal Government customers only.
Synonym: Coulometric Single Solution
Standard, Water, 0.1%, Apura™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Standard, Water Name Note: Apura™ Karl Fischer Water Standard; 0.1mg H2O/g Assay Percent Range: 0.10%
Standard, Water, 1%, Apura™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Standard, Water Name Note: Apura™ Karl Fischer Water Standard, Mixture. Contains 1-Methoxy-2-propanol. Assay Percent Range: 1.00%
Standard, Water, 0.01%, Apura™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Standard, Water Name Note: Apura™ Karl Fischer Water Standard; 0.1mg H2O/g Assay Percent Range: 0.01%
CombiCoulomat Frit Reagent, AQUASTAR™ EMD Chemicals Available on GSA/VA Contract for Federal Government customers only.
Synonym: Coulometric Single Solution
AquaStar Coulomat A (Original Formulation), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.809 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N Synonym: Coulometric Anode Solution PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: molecular iodine SMILES: II
HYDRANAL™ - Medium K, Honeywell Fluka™ Available on GSA/VA Contract for Federal Government customers only.
Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free).
AQUASTAR™ Composite 2K (Original Formulation), EMD Millipore
CAS: 7553-56-2 Molecular Formula: I2 Molecular Weight (g/mol): 253.809 InChI Key: PNDPGZBMCMUPRI-UHFFFAOYSA-N PubChem CID: 807 ChEBI: CHEBI:17606 IUPAC Name: molecular iodine SMILES: II
HYDRANAL™ - Buffer Acid, Buffer substance for KF titration, Honeywell Fluka™ Available on GSA/VA Contract for Federal Government customers only.
Liquid buffer medium for Karl Fischer titration in acidic samples (based on imidazole).
  spinner