Sodium Hydroxide
Sodium Hydroxide Solution (2.5N/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution, 2.5N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: NaOH · H2O MDL Number: MFCD00003548
Sodium Hydroxide (Pellets/NF/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: NF/FCC Physical Form: Solid Assay: 95.0 to 100.5% (FCC) Molecular Formula: NaOH Formula Weight: 40g/mol MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium Hydroxide Solution (50% w/w/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 50% (w/w) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Assay: Inclusive between 50% MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide (Beads) 2, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Physical Form: Solid Molecular Formula: NaOH Formula Weight: 40g/mol MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium Hydroxide (NF/EP/BP/FCC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: NF/EP/BP/FCC Assay: 95.0 to 100.5% (FCC) Molecular Formula: NaOH Formula Weight: 40g/mol MDL Number: MFCD00003548
Sodium Hydroxide Solution (1N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 1N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide (Granular/NF/FCC/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 Purity Grade: NF/FCC/EP/BP/JP Physical Form: Solid Assay: 95.0 to 100.5% (NF) Molecular Formula: NaOH Formula Weight: 40.00g/mol MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Purity Grade: Certified ACS Physical Form: Solid Assay: 97.0 % min. Assay Percent Range: ≥97.0 % Molecular Formula: NaOH Formula Weight: 40g/mol MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium Hydroxide 1N Concentrate, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide 1N Concentrate Name Note: For dilution to 0.1N CAS: 1310-73-2 CAS: 7732-18-5 Physical Form: Liquid MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.25N (N/4)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.25N (N/4) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: HNaO MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (50% w/w), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 50% (w/w) CAS: 1310-73-2 CAS: 7732-18-5 Assay: Inclusive between 50% MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.5N (N/2)/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.5N (N/2) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.02N (N/50)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.02N (N/50) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.01N (N/100)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.01N (N/100) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.2N (N/5)/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.2N (N/5) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: HNaO MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution, 2% (AOAC), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution, 2% CAS: 7732-18-5 CAS Min %: 94.0 CAS Max %: 98.0 CAS: 1310-73-2 CAS Min %: 3.79 CAS Max %: 3.94
Sodium Hydroxide Solution (10N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 10N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: NaOH MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution, 40%, Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 40% (w/w) CAS: 1310-73-2 CAS: 7732-18-5 Assay: 40 to 42% MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide (Beads) 1, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 CAS: 497-19-8 Physical Form: Solid Molecular Formula: NaOH Formula Weight: 40g/mol MDL Number: MFCD00003548 Synonym: Caustic Soda
Sodium Hydroxide Solution (2N/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 2N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.1N (N/10)/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.1N (N/10) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (0.05N (N/20)/Certified), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 0.05N (N/20) CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution (6.25N), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 6.25N CAS: 1310-73-2 CAS: 7732-18-5 Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide 10N Concentrate (Certified) ACS, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide 10N Concentrate CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified ACS Physical Form: Liquid Molecular Formula: NaOH MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide Solution, 2.5N (AOAC), Fisher Chemical
Chemical Name or Material: Sodium Hydroxide Solution, 2.5N CAS: 7732-18-5 CAS Min %: 82.0 CAS Max %: 85.0 CAS: 1310-73-2 CAS Min %: 16.39 CAS Max %: 17.05
Sodium Hydroxide Solution (5N/Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 5N CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Molecular Formula: NaOH · H2O MDL Number: MFCD00003548 Synonym: Caustic Soda Solution
Sodium Hydroxide, 33.3% (w/w) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Sodium Hydroxide CAS: 7732-18-5 CAS Min %: 67.0 CAS Max %: 70.0 CAS: 1310-73-2 CAS Min %: 31.03 CAS Max %: 32.29 Physical Form: Clear colorless liquid
Sodium Hydroxide, 40% (w/v) Aqueous Solution, Nitrogen Free, Suitable for Kjeldahl Nitrogen, Ricca Chemical
Chemical Name or Material: Sodium Hydroxide Solution Name Note: 40% (w/v), Nitrogen Free
Sodium Hydroxide, 0.100 Normal (N/10), Ricca Chemical
Chemical Name or Material: Sodium Hydroxide CAS: 7732-18-5 CAS Min %: 97.0 CAS Max %: 100.0 CAS: 1310-73-2 CAS Min %: 0.78 CAS Max %: 0.81 Physical Form: Clear colorless liquid
Sodium Hydroxide ACS AR Pellet, Macron Fine Chemicals™
Chemical Name or Material: Sodium Hydroxide CAS: 1310-73-2 Purity Grade: ACS Physical Form: Pellets Molecular Formula: NaOH Formula Weight: 40g/mol
  spinner