Life Science Buffers
Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: Laboratory Physical Form: Powder; Solid Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14g/mol MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
Tris (Tromethamine) (Granular/USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris (Tromethamine) CAS: 77-86-1 Purity Grade: USP Physical Form: Powder; Solid Assay: 99.0 to 101.0% (USP) Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14g/mol MDL Number: MFCD00004679 Synonym: THAM™
Tris SDS PAGE Running Buffer, FASTRun, 10X
CAS: 151-21-3 CAS: 151-21-3 CAS: 7732-18-5 Purity Grade: Protein Electrophoresis Synonym: Denaturing Buffer
Phosphate Buffer, 0.05M, Fisher Chemical
Chemical Name or Material: Phosphate Buffer, 0.05M CAS: 7732-18-5 CAS Min %: 91.0 CAS Max %: 95.0 CAS: 64-19-7 CAS Min %: 5.76 CAS Max %: 5.99 CAS: 7778-77-0 CAS Min %: 0.31 CAS Max %: 0.32 CAS: 7664-38-2 CAS Min %: 0.26 CAS Max %: 0.27 CAS: 6153-56-6 CAS Min %: 0.18 CAS Max %: 0.19 CAS: 79-09-4 CAS Min %: 0.09 CAS Max %: 0.09
Tris(hydroxymethyl)aminomethane Hydrochloride (Crystalline/Certified) ACS, Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane Hydrochloride CAS: 1185-53-1 Purity Grade: Certified ACS Physical Form: Solid Assay: 95% min. Assay Percent Range: ≥95 % Molecular Formula: C4H11NO3 · HCl Linear Formula: NH2C(CH2OH)3·HCl Formula Weight: 157.6g/mol MDL Number: MFCD00012590 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol Hydrochloride
Tris(hydroxymethyl)aminomethane (Cryst./Certified ACS), Fisher Chemical
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: Certified ACS Physical Form: Powder; Solid Assay: 99.8 to 100.1% Assay Percent Range: 99.8 to 100.1% Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14g/mol MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Acetate-EDTA Name Note: 10X Solution CAS: 64-19-7 CAS: 7732-18-5 CAS: 60-00-4 CAS: 77-86-1 Purity Grade: Electrophoresis Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: TAE
Tween™ 80, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 80 CAS: 9005-65-6 Physical Form: Liquid Synonym: Polyoxyethylene-20-sorbitan Monooleate
TBE Buffer, Tris-Borate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Chemical Name or Material: Tris-Borate-EDTA Name Note: 1X Solution CAS: 77-86-1 CAS: 6381-92-6 CAS: 10043-35-3 CAS: 7732-18-5 Purity Grade: Electrophoresis
TE Buffer, Tris-EDTA, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Chemical Name or Material: Tris-EDTA Name Note: 1X Solution, pH 7.4 CAS: 77-86-1 CAS: 60-00-4 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Synonym: TE
TAE Buffer, Tris-Acetate-EDTA, 1X Solution, Electrophoresis, Fisher BioReagents
Chemical Name or Material: Tris-Acetate-EDTA Name Note: 1X Solution CAS: 77-86-1 CAS: 6850-28-8 CAS: 139-33-3 CAS: 10378-23-1 CAS: 7732-18-5 Purity Grade: Electrophoresis
TE Buffer, Tris-EDTA, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Chemical Name or Material: Tris-EDTA (TE), 10X Solution Name Note: pH 7.4 CAS: 77-86-1 CAS: 6381-92-6 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Synonym: TE
Tween™ 20, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tween™ 20 CAS: 9005-64-5 Physical Form: Viscous Liquid Molecular Formula: C58H114O26 Formula Weight: 1227.54g/mol Synonym: Polyoxyethylene-20-sorbitan Monolaurate
TBE Buffer, Tris-Borate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Borate-EDTA Name Note: 10X Solution CAS: 10043-35-3 CAS: 60-00-4 CAS: 77-86-1 CAS: 7732-18-5 Purity Grade: Electrophoresis Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: TBE
Tris-Glycine, 10X Solution (Electrophoresis), Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Glycine Name Note: 10X Solution CAS: 7732-18-5 CAS: 77-86-1 CAS: 56-40-6 Purity Grade: Electrophoresis Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: TG
Tris-EDTA, 100X Solution (Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris-EDTA Name Note: 100X Solution CAS: 77-86-1 CAS: 60-00-4 CAS: 7732-18-5 Purity Grade: Molecular Biology
Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris Hydrochloride Name Note: 1M Solution, pH 8.0 CAS: 77-86-1 CAS: 1185-53-1 CAS: 7732-18-5 Purity Grade: Molecular Biology Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: Tris-HCl
TBE Buffer, 10X Powder, Electrophoresis, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Borate-EDTA Name Note: 10X Powder CAS: 77-86-1 CAS: 10043-35-3 CAS: 60-00-4 Purity Grade: Electrophoresis
SDS-PAGE Buffer, 5X Powder, Electrophoresis, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Glycine-SDS CAS: 56-40-6 CAS: 77-86-1 CAS: 151-21-3 Purity Grade: Electrophoresis Physical Form: Solid Synonym: TGS Buffer
Tris Buffer, 0.3M Solution (Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris Buffer Name Note: 0.3M Solution CAS: 7732-18-5 CAS: 77-86-1 CAS: 60-00-4 Purity Grade: Molecular Biology
Tris-Glycine Buffer, 10X Powder, Electrophoresis, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Tris-Glycine Name Note: 10X Powder CAS: 77-86-1 CAS: 56-40-6 Purity Grade: Electrophoresis
Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents
Chemical Name or Material: Tris Hydrochloride Name Note: 1M Solution, pH 7.0 CAS: 77-86-1 CAS: 1185-53-1 CAS: 7732-18-5 Purity Grade: Molecular Biology Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Synonym: Tris-HCl
Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent, ACROS Organics™
Chemical Name or Material: Tris(hydroxymethyl)aminomethane CAS: 77-86-1 Purity Grade: ACS Reagent Assay: 99.8 to 100.1% (on dry basis) Assay Percent Range: 99.8% Molecular Formula: C4H11NO3 Linear Formula: (HOCH2)3CNH2 Formula Weight: 121.14g/mol MDL Number: MFCD00004679 Synonym: 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
Tris Buffer, 2M Solution (Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris Buffer Name Note: 2M Solution CAS: 77-86-1 Purity Grade: Molecular Biology
Bis-Tris, Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Bis-Tris CAS: 6976-37-0 Physical Form: Powder; Solid Assay: 99% min. Assay Percent Range: ≥99 % Molecular Formula: C8H19NO5 Linear Formula: (HOCH2CH2)2NC(CH2OH)3 Formula Weight: 209.24g/mol Synonym: 2, 2-Bis(hydroxymethyl)-2, 2?
Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents
Chemical Name or Material: Tris Base CAS: 77-86-1 Purity Grade: Molecular Biology Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Powder; Solid Assay: 99.8% min. Assay Percent Range: ≥99.8 % Molecular Formula: C4H11NO3 Linear Formula: H2NC(CH2OH)3 Formula Weight: 121.14g/mol Synonym: Trimethylol Aminomethane
STET Buffer, 1X Solution (pH 8.0), Fisher BioReagents
Chemical Name or Material: STET Buffer, 1X Solution Name Note: pH 8.0 CAS: 77-86-1 (Tris Base) CAS: 60-00-4 (EDTA) CAS: 7647-01-0 (HCl) CAS: 7647-14-5 (NaCl) CAS: 9002-93-1 (Triton™ X-100)
TE Buffer, Tris-EDTA, 1X Solution, pH 7.6, Molecular Biology, Fisher BioReagents
Chemical Name or Material: Tris-EDTA Name Note: 1X Solution, pH 7.6 CAS: 77-86-1 CAS: 6381-92-6 CAS: 7647-01-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Synonym: TE
Protein Gel-Loading Dye for SDS-PAGE, 2X (Dark-Purple Liq./Electrophor.), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Protein Gel-Loading Dye for SDS-PAGE, 2X CAS: 7732-18-5 CAS: 56-81-5 CAS: 77-86-1 CAS: 151-21-3 CAS: 7647-01-0 Purity Grade: Electrophoresis Physical Form: Liquid
TAPS, 99+%, for biochemistry, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: TAPS CAS: 29915-38-6 Purity Grade: For biochemistry Assay: 99% min. Assay Percent Range: 99+% Molecular Formula: C7H17NO6S Linear Formula: (HOCH2)3CNH(CH2)3SO3H Formula Weight: 243.27g/mol MDL Number: MFCD00007538 Synonym: 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
  spinner