pH Reference Buffers
Buffer Solution, pH 10.00, Color-Coded Blue (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 10.00, Color-Coded Blue CAS: 7732-18-5 CAS: 6381-92-6 CAS: 584-08-7 CAS: 1332-77-0 CAS: 1310-58-3 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 2.00 (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 2.00 CAS: 7447-40-7 CAS: 7647-01-0 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 10.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 10.00 CAS: 6381-92-6 CAS: 584-08-7 CAS: 1310-58-3 CAS: 7732-18-5 CAS: 12228-88-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 10, Color-coded blue, Certified, ACROS Organics™
Chemical Name or Material: Buffer solution, pH 10.00 Name Note: Traceable to NIST CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD01324186
Buffer Solution, pH 6.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 6.00 CAS: 7778-77-0 CAS: 7732-18-5 CAS: 1310-73-2 Purity Grade: Certified Grade MDL Number: MFCD00134130
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use, ACROS Organics™
Chemical Name or Material: Potassium hydrogen phthalate CAS: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.22 MDL Number: MFCD00217672
Buffer solution, pH 4.00, color-coded red Certified, traceable to NIST, ACROS Organics™
Chemical Name or Material: Buffer solution, pH 4.00 Name Note: Color-coded Red CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer solution pH 9.2, Borate buffer, traceable to NIST, ready to use, ACROS Organics™
Chemical Name or Material: Buffer solution pH 9.2 Name Note: Borate buffer, traceable to NIST, ready to use CAS: 1330-43-4 MDL Number: MFCD00163147
Buffer Concentrate, pH 7.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Concentrate, pH 7.00 CAS: 64-02-8 CAS: 7732-18-5 CAS: 1310-58-3 CAS: 1310-73-2 CAS: 7664-38-2 Purity Grade: Certified Grade Physical Form: Liquid MDL Number: MFCD00146206
Buffer Solution, pH 4.63 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 4.64 CAS: 64-19-7 CAS: 6131-90-4 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00137248
Buffer solution pH 4, Phthalate buffer, traceable to NIST, colored red, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer solution pH 4 Name Note: Phthalate buffer, traceable to NIST CAS: 7732-18-5 CAS: 7487-94-7 CAS: 3244-88-0 MDL Number: MFCD00217672
Buffer Solution, pH 7.00 (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 7.00 CAS: 7778-77-0 CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00134130 Synonym: Chlorine Titrimeter Reagent
Buffer Solution, pH 5.00 (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 5.00 CAS: 877-24-7 CAS: 1310-73-2 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 4.00 (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 4.00 CAS: 877-24-7 CAS: 7732-18-5 CAS: 50-00-0 CAS: 67-56-1 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 7.40 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 7.40 CAS: 7778-77-0 CAS: 64-02-8 CAS: 7732-18-5 CAS: 7758-11-4 Purity Grade: Certified Grade MDL Number: MFCD00134130
Buffer-Pac Color-Coded Solutions (Certified), Fisher Chemical
Chemical Name or Material: BufferPac™ Color-Coded Solutions CAS: 50-00-0 Purity Grade: Certified Grade MDL Number: MFCD00163177
Buffer Solution, pH 9.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 9.00 CAS: 7447-40-7 CAS: 10043-35-3 CAS: 1310-73-2 CAS: 64-02-8 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer solution pH 4, Citrate buffer, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer solution pH 4 Name Note: Phthalate buffer CAS: 877-24-7 CAS: 7647-14-5 CAS: 68-04-2 CAS: 7732-18-5 Molecular Formula: C8H5KO4 Formula Weight: 204.23 MDL Number: MFCD00130806
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use, ACROS Organics™
Chemical Name or Material: Buffer solution pH 7 Name Note: Phosphate buffer, traceable to NIST, ready to use CAS: 7558-79-4 MDL Number: MFCD03097944
Buffer solution, pH 7.00 , Color-coded yellow, Certified, ACROS Organics™
Chemical Name or Material: Buffer solution pH 7.00 Name Note: Color-coded yellow CAS: 7778-77-0 MDL Number: MFCD00146206
Buffer Concentrate, pH 11.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Concentrate, pH 11.01 CAS: 56-40-6 CAS: 7647-14-5 CAS: 1310-73-2 CAS: 139-33-3 CAS: 7732-18-5 Purity Grade: Certified Grade Physical Form: Liquid MDL Number: MFCD00163176
Buffer Solution, pH 10, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer solution pH 10 CAS: 1332-57-6 CAS: 144-55-8 CAS: 7732-18-5
Buffer Solution, pH 4.00, Color-Coded Red (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 4.00, Color-Coded Red CAS: 50-00-0 CAS: 67-56-1 CAS: 877-24-7 CAS: 7732-18-5 CAS: 16423-68-0 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Solution, pH 3.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Solution, pH 3.00 CAS: 877-24-7 CAS: 7647-01-0 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer Concentrate, pH 10.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Concentrate, pH 10.01 CAS: 7447-40-7 CAS: 1310-73-2 CAS: 139-33-3 CAS: 7732-18-5 CAS: 10043-35-3 Purity Grade: Certified Grade Physical Form: Liquid MDL Number: MFCD00163176
Buffer solution pH 5, Acetate buffer, ACROS Organics™
Chemical Name or Material: Buffer solution pH 5 CAS: 77-92-9 CAS: 7732-18-5 CAS: 64-19-7 CAS: 1310-58-3 MDL Number: MFCD03097943
Buffer Concentrate, pH 4.00 (Certified), Fisher Chemical
Chemical Name or Material: Buffer Concentrate, pH 4.00 CAS: 877-24-7 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade Physical Form: Liquid MDL Number: MFCD00013070
Buffer Solution, pH 1.00 (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 1.01 CAS: 7447-40-7 CAS: 7647-01-0 CAS: 50-00-0 CAS: 67-56-1 CAS: 7732-18-5 Purity Grade: Certified Grade MDL Number: MFCD00146206
Buffer solution pH 10, Borate buffer, traceable to NIST, ready to use, ACROS Organics™
Chemical Name or Material: Buffer solution pH 10 Name Note: Borate buffer, traceable to NIST, ready to use MDL Number: MFCD00163147
Buffer Solution, pH 7.00, Color-Coded Yellow (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Buffer Solution, pH 7.00, Color-Coded Yellow CAS: 7778-77-0 CAS: 1310-73-2 CAS: 7732-18-5 CAS: 1934-21-0 Purity Grade: Certified Grade MDL Number: MFCD00134130
  spinner