Application Specific Chloroform
Chloroform (Approx. 0.75% Ethanol as Preservative/HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 0.75% Ethanol as Preservative CAS: 67-66-3 CAS: 64-17-5 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.8% Assay Percent Range: 99.8% min by GC Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform (Amylene as Preservative, Pesticide), Fisher Chemical
Chemical Name or Material: Chloroform Name Note: With 150-200ppm amylene as preservative CAS: 67-66-3 CAS: 513-35-9 Purity Grade: Pesticide Grade Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 50ppm Pentene as Preservative CAS: 67-66-3 CAS: 513-35-9 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.8% Assay Percent Range: 99.8% min by GC Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform (Ethanol as Preservative, Spectranalyzed™), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 0.75% ethanol as preservative CAS: 67-66-3 CAS: 64-17-5 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform, with amylene as preservative, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 50ppm Amylene as Preservative CAS: 67-66-3 CAS: 513-35-9 Purity Grade: OPTIMA™ Purity Grade Notes: High purity mobile phase for HPLC and/or extraction solvent for GC applications Assay: ≥99.9% Assay Percent Range: 99.9% min by GC Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 Synonym: Trichloromethane
Chloroform, 99+%, for spectroscopy, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Stabilized with amylene CAS: 67-66-3 CAS: 513-35-9 Purity Grade: For spectroscopy Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Alfa Aesar™ Chloroform, ethanol-free, 99+%, stab. with ca 50 ppm amylene
Chemical Name or Material: Chloroform CAS: 67-66-3 Name Note: ethanol-free, stabilized With ≈50ppm amylene Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 UN Number: UN1888 Beilstein: 1731042 Merck Index: 14,2141 Synonym: Trichloromethane
Chloroform (Approx. 0.75% Ethanol as Preservative/Molecular Biology), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Approx. 0.75% Ethanol as Preservative CAS: 67-66-3 CAS: 64-17-5 Purity Grade: Molecular Biology Grade Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % (by GC) Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, Spectrophotometric Grade, greater-than99.5%, Alfa Aesar™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Alfa Aesar™ Chloroform, 99%, stab. with 0.8-1% ethanol
Chemical Name or Material: Chloroform CAS: 67-66-3 Name Note: stabilized with 0.8 to 1% ethanol Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826 UN Number: UN1888 Beilstein: 1731042 Merck Index: 14,2141 Synonym: Trichloromethane
Chloroform ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Chloroform CAS: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, 99.9%, for residue analysis, ECD tested for pest anal., residue <0., ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: ECD Tested, For Pesticide Analysis, Residue <0.0005% CAS: 67-66-3 CAS: 513-35-9 Purity Grade: For residue analysis Assay: 99.9% Assay Percent Range: 99.9% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, HPLC Grade, Alfa Aesar™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
ChloroForm, HPLC, J.T.Baker™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: HPLC Grade Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, HPLC Grade, >99.5% min, Alfa Aesar™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
ChloroForm, HPLC, J.T.Baker™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: HPLC Grade Purity Grade Notes: Meets ACS reagent chemical requirements Molecular Formula: CHCl3 Formula Weight: 119.38
Chloroform, OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Formula Weight: 119.38
Chloroform, stabilized with ethanol, for HPLC, ACROS Organics™
Chemical Name or Material: Chloroform Name Note: Stabilized with ethanol CAS: 67-66-3 CAS: 64-17-5 Purity Grade: For HPLC Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, for HPLC, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: Stabilized CAS: 67-66-3 CAS: 513-35-9 Purity Grade: For HPLC Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, 99.9% min., OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Physical Form: Liquid Assay Percent Range: 99.9% min. Molecular Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene, 1% ethanol, >99.9% (excluding ethanol), Honeywell Burdick & Jackson
CAS Max %: 1.0000% Density: 1.49 g/cm3 Boiling Point: 62°C Solubility: 81.5% in Water at 20°C Formula Weight: 119.37 Purity Grade: for HPLC Water: 0.02% max. Absorbance: 0.3 AU max. at 250 nm (UV) Solubility Information: 81.5% in Water at 20°C CAS: 67-66-3 CAS: 513-35-9 CAS: 64-17-5 Synonym: TRICHLOROMETHANE Chemical Name or Material: Chloroform CAS Max %: 98.9900% Beilstein: 1731042 Name Note: B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene and 1% ethanol, >99.9% (excluding ethanol) Packaging: Glass Bottle Assay: >99.9% CAS Max %: <0.0100% UN Number: UN1888
Chloroform, HPLC, 99.8% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Name Note: For ACS analysis Purity Grade: ACS/HPLC/NF/USP Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 Synonym: Trichloromethane
Chloroform, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Linear Formula: CHCl3 Formula Weight: 119.38
Chloroform, For gas chromatography ECD and FID SupraSolv™ EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform CAS: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38 UN Number: 1888 Synonym: TCM, Trichloromethane, Methane trichloride
Chloroform, for HPLC, stabilized with amylene, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Chloroform Name Note: 99.8% CAS: 67-66-3 CAS Min %: 99.0 CAS: 513-35-9 CAS Min %: 1.0 Purity Grade: For HPLC Assay: ≥99.8% Molecular Formula: CHCl3 Linear Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
Chloroform, Environmental Grade, 99.8+%, stab. with ethanol, Alfa Aesar™
Chemical Name or Material: Chloroform CAS: 67-66-3 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >99.8% Molecular Formula: CHCl3 Formula Weight: 119.38 MDL Number: MFCD00000826
  spinner