Denatured Alcohols
Reagent Alcohol, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Reagent Alcohol CAS: 64-17-5 CAS: 67-63-0 CAS: 67-56-1 Purity Grade: Histological Grade Purity Grade Notes: Reagent Physical Form: Liquid Assay Percent Range: Assay: Ethanol (88 to 91%), Methanol (4.0 to 5.0% v/v), Isopropyl Alcohol (4.5 to 5.5%) MDL Number: MFCD00003568
Alcohol (HPLC), Fisher Chemical
Chemical Name or Material: Alcohol CAS: 67-56-1 CAS: 64-17-5 CAS: 67-63-0 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Molecular Formula: CH3CH2OH Linear Formula: CH3CH2OH MDL Number: MFCD00003568 Synonym: Ethanol, Ethyl Alcohol, Denatured Alcohol
Ethyl Alcohol Denatured (Proprietary Solvent), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol Denatured Name Note: Proprietary Solvent CAS: 64-17-5 MDL Number: MFCD00003568 Synonym: Ethanol, Denatured
Alcohol, Reagent (Denatured Alcohol), 70% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Alcohol, Reagent Name Note: Denatured Alcohol CAS: 64-17-5 CAS Min %: 62.0 CAS Max %: 65.0 CAS: 7732-18-5 CAS Min %: 29.4 CAS Max %: 30.6 CAS: 67-63-0 CAS Min %: 3.43 CAS Max %: 3.57 CAS: 67-56-1 CAS Min %: 3.09 CAS Max %: 3.21 Physical Form: Liquid
Ethanol, for HPLC, denatured, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: For HPLC, Denatured CAS: 64-17-5 CAS: 67-56-1 CAS: 67-63-0 Purity Grade: For HPLC Molecular Formula: C2H6O Linear Formula: C2H5OH Formula Weight: 46.06 MDL Number: MFCD00003568 Synonym: Ethyl alcohol
Specially Denatured Ethanol SDA 3A, Decon Laboratories Inc
Chemical Name or Material: Denatured Ethanol CAS: 64-17-5 CAS: 67-56-1 Physical Form: Liquid Synonym: Ethyl Alcohol
Ethanol, CDA 19 (Histological), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol, CDA 19 CAS: 64-17-5 CAS: 108-10-1 CAS: 108-88-3 CAS: 110-54-3 CAS: 7732-18-5 Purity Grade: Histological Grade Physical Form: Liquid MDL Number: MFCD00003568 Synonym: Ethyl Alcohol Denatured
SDA 3-A Ethanol, 190 Proof (Denatured), Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: 190 Proof CAS: 64-17-5
Alcohol, Reagent , ACS Reagent Grade, Anhydrous, Absolute, Ricca Chemical
Chemical Name or Material: Alcohol, Reagent , Anhyd., Absolute Name Note: Denatured alcohol CAS: 64-17-5 CAS Min %: 90.0 CAS Max %: 91.0 CAS: 67-63-0 CAS Min %: 4.0 CAS Max %: 6.0 CAS: 67-56-1 CAS Min %: 4.0 CAS Max %: 5.0 Purity Grade: ACS Reagent Grade
Reagent Alcohol, Certified, 70% (v/v), LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Reagent Alcohol CAS: 64-17-5 CAS Max %: 56.46 CAS: 7732-18-5 CAS Max %: 37.63 CAS: 67-63-0 CAS Max %: 3.11 CAS: 67-56-1 CAS Max %: 2.8 Purity Grade: Certified Grade Linear Formula: C2H6O Formula Weight: 46.07 UN Number: UN1987 Synonym: Ethyl Alcohol; SDA 3A
Ethanol, Alcohol Reagent, anhydrous, denatured, ACS, 94 to 96%, Alfa Aesar™
Chemical Name or Material: Ethanol, Alcohol Reagent, anhydrous, denatured Name Note: For ACS analysis; Begins to lose water at 60°, anhydrous at 100° CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 94-96% Molecular Formula: C2H6O Formula Weight: 46.08 MDL Number: MFCD00003568
Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Alcohol Name Note: Anhydrous Purity Grade: Baker Analyzed™ ACS Reagent Purity Grade Notes: Meets ACS reagent chemical requirements
Reagent Alcohol ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Reagent Alcohol Name Note: Absolute CAS: 64-17-5 Purity Grade: ACS Reagent Grade Molecular Formula: CH3CH2OH Formula Weight: 46.07
Ethanol, SDA 40B, 200 Proof, PHARMCO-AAPER™
CAS: 64-17-5 CAS: 75-65-0 CAS: 3734-33-6 Physical Form: Liquid, Invisible Vapor Molecular Formula: C2H6O Formula Weight: 46.07 Synonym: Denatured Alcohol 40B, Denatured ethanol, Ethanol denatured with tert-butyl alcohol and denatonium benzoate
Alcohol, OmniSolv™ Reagent (HPLC), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Components: 90% (w/v) Ethyl Alcohol (CAS Reg. 64-17-5) + 5% (w/v) Methanol (CAS Reg. 67-56-1) + 5% (w/v) 2-Propanol (CAS Reg. 67-63-0)
Alcohol Reagent ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Alcohol Name Note: Reagent CAS: 67-63-0 Purity Grade: ACS Reagent Grade Linear Formula: CH3CH2OH with methanol and isopropyl alcohol Formula Weight: 46.07
Ethyl Alcohol, Reagent, Denat., Spectro Grade, Contains 5% 2-PrOH and 5% MeOH, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol CAS: 64-17-5 CAS: 67-56-1 CAS: 67-63-0 Purity Grade: For spectroscopy Assay: 4.0 to 6.0% (2-propanol) Molecular Formula: C2H6O Linear Formula: C2H5OH Formula Weight: 46.06
Alcohol, Reagent, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Alcohol, Reagent Physical Form: Liquid Linear Formula: CHCH3CHCH2OH Formula Weight: 46.7 Synonym: Ethyl Alcohol, Denatured
Ethyl Alcohol, 95% Denatured Special, 84.0 to 86.0%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol Name Note: Denatured Special CAS: 64-17-5 Physical Form: Liquid Assay Percent Range: 95% Molecular Formula: CH3CH2OH Linear Formula: CH3CH2OH + denaturants Formula Weight: 46.7 Synonym: Ethanol
Alcohol, Reagent, 20% (v/v), Fisher Chemical
Chemical Name or Material: Reagent Alcohol, 20% (v/v) CAS: 7732-18-5 CAS Min %: 79.0 CAS Max %: 81.0 CAS: 64-17-5 CAS Min %: 18.0 CAS Max %: 20.0 CAS: 67-56-1 CAS Min %: 0.8 CAS Max %: 0.9 CAS: 67-63-0 CAS Min %: 0.9 CAS Max %: 1.0
Ethanol, anhydrous, denatured, HPLC Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™
Chemical Name or Material: Ethanol CAS: 64-17-5 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 90% Molecular Formula: C2H6O Formula Weight: 46.08 MDL Number: MFCD00003568
Ethyl Alcohol Anyhydrous (Denatured), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Anyhydrous Ethyl Alcohol (Denatured) Formula Weight: 46.07
Alcohol, Reagent, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Alcohol, Reagent CAS: 7732-18-5 CAS Min %: 49.0 CAS Max %: 51.0 CAS: 64-17-5 CAS Min %: 45.0 CAS Max %: 46.0 CAS: 67-63-0 CAS Min %: 2.0 CAS Max %: 3.0 CAS: 67-56-1 CAS Min %: 2.0 CAS Max %: 3.0 Physical Form: Liquid
Ethanol, anhydrous, denatured, Spectrophotometric Grade, 90%, 5% methanol, 5% isopropanol, Alfa Aesar™
Chemical Name or Material: Ethanol CAS: 64-17-5 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 90% Molecular Formula: C2H6O Formula Weight: 46.08 MDL Number: MFCD00003568
Ethanol Solution, Denatured, Sterile, Made with USP Water for Injection, 70%, BAKER™, J.T.Baker™
Chemical Name or Material: Ethanol Solution Name Note: Denatured, Sterile, Made with USP Water for Injection Formula Weight: 46.07
Reagent Alcohol, ACS Grade, 94.0% to 96.0% (v/v), LabChem™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Reagent Alcohol CAS: 64-17-5 CAS Max %: 90.25 CAS: 67-63-0 CAS Max %: 5 CAS: 67-56-1 CAS Max %: 4.75 Purity Grade: ACS Grade Linear Formula: C2H6O Formula Weight: 46.07 UN Number: UN1987 Synonym: Ethyl Alcohol; SDA 3A
Ethyl Alcohol Anhydrous, Denatured Special, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol Name Note: Denatured Special Physical Form: Liquid Linear Formula: CH3CH2OH + denaturants Formula Weight: 46.7
Alcohol, Anhydrous, Contains 0.075% Butylated Hydroxytoluene as a stabilizer, PHOTREX™ Reagent, J.T.Baker™
Chemical Name or Material: Alcohol Name Note: Anhydrous, Contains 0.075% Butylated Hydroxytoluene as a stabilizer Purity Grade: PHOTREX™ Reagent Purity Grade Notes: Meets ACS reagent chemical requirements
Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK
Chemical Name or Material: Ethanol CAS: 64-17-5 Name Note: Anhydrous, denatured Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.08 MDL Number: MFCD00003568 UN Number: UN1993 Beilstein: 1718733 Merck Index: 14,3760 Synonym: Ethyl alcohol
Alcohol, Anhydrous, Reagent, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Alcohol Name Note: Anhydrous, Reagent Purity Grade: Baker Analyzed™ ACS Reagent
  spinner