Application Specific Ethanol
Ethanol, 200 proof, Molecular Biology Grade, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: 200 Proof (100% ethanol) CAS: 64-17-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: Made to USP specifications, Tested for the presence of DNase and Rnase Assay Percent Range: ≥99.5% Synonym: Ethyl Alcohol
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: Absolute (200 Proof) CAS: 64-17-5 Purity Grade: Molecular Biology Grade Purity Grade Notes: DNase-, RNase- and Protease-Free Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: Assay: ≥99.5% (v/v) Molecular Formula: C2H6O Linear Formula: C2H5OH Formula Weight: 46.07 Synonym: Ethyl Alcohol
Ethanol Solution 96%, Molecular Biology Grade, Regulated, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 96% Ethanol Solution Purity Grade: Molecular Biology Grade Synonym: 96% Ethyl Alcohol Solution CAS: 64-17-5 CAS: 7732-18-5
Ethanol, 70% Solution, Molecular Biology Grade, Denatured, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 70% Denatured Ethanol Solution (Molecular Biology Grade) CAS: 64-17-5 CAS: 67-63-0 CAS: 7732-18-5 Purity Grade: Molecular Biology Grade Synonym: 70% Denatured Ethyl Alcohol Solution
Ethyl Alcohol Non UV, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethyl Alcohol Name Note: Non UV, Denatured CAS: 64-17-5 Physical Form: Liquid Linear Formula: CH3CH2OH + denaturants Formula Weight: 46.7 Synonym: Ethanol
Ethyl Alcohol, Pure, 200 Proof, OmniPur™, EMD Millipore
Chemical Name or Material: Ethyl Alcohol Molecular Formula: C2H6O Formula Weight: 46.08 UN Number: 1170 Synonym: Ethanol, Alcohol
Ethanol Solution 70%, Molecular Biology Grade, Fisher BioReagents™ Available on GSA/VA Contract for Federal Government customers only.
Physical Form: Liquid Chemical Name or Material: 70% Ethanol Solution Purity Grade: Molecular Biology Grade Ethanol: 68%-72% (GC-TCD) Water: 28%-32% (GC-TCD) Color: Clear Colorless RNase: Not Detected DNase: Not Detected Protease: Not Detected
Alfa Aesar™ Ethanol, denatured, 91.6%, 3.7% methanol, 1.9% MIBK, 1% heptane, 1% ethyl acetate, 1% toluene (v/v)
Chemical Name or Material: Ethanol CAS: 64-17-5 Name Note: Denatured Formula Weight: 46.08 MDL Number: MFCD00003568 UN Number: UN1170 Beilstein: 1718733 Merck Index: 14,3760 Synonym: Ethyl alcohol
Alfa Aesar™ Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK
Chemical Name or Material: Ethanol CAS: 64-17-5 Name Note: Anhydrous, denatured Molecular Formula: C2H6O Linear Formula: CH3CH2OH Formula Weight: 46.08 MDL Number: MFCD00003568 UN Number: UN1993 Beilstein: 1718733 Merck Index: 14,3760 Synonym: Ethyl alcohol
Ethanol, denatured, Spectrophotometric Grade, 90%, Alfa Aesar™
Chemical Name or Material: Ethanol, denatured CAS: 64-17-5 CAS: 67-56-1 CAS: 67-63-0 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 90% Molecular Formula: C2H6O Formula Weight: 46.08 MDL Number: MFCD00003568
Reagent Alcohol, 18.5% (v/v) Denatured Alcohol, Ricca Chemical
Chemical Name or Material: Reagent Alcohol Name Note: Denatured Alcohol Assay Percent Range: 18.5% (v/v)
Reagent Alcohol, 13.5% (v/v) Denatured Alcohol, Ricca Chemical
Chemical Name or Material: Reagent Alcohol Name Note: Denatured Alcohol Assay Percent Range: 13.5% (v/v)
Ethanol for Spectroscopy Uvasol™ EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol CAS: 64-17-5 Molecular Formula: C2H6O Formula Weight: 46.07 UN Number: 1170 Synonym: EtOH
  spinner