Multicompendial Ethanol
Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical
Benzene: 2ppm max. (USP, EP) Boiling Point: 78°C CAS: 64-17-5 Chemical Name or Material: Ethanol, 200 Proof Color: Clear Formula Weight: 46.07 Melting Point: −114°C Methanol: 200ppm (USP, EP) Physical Form: Liquid Purity Grade: EP/USP/ACS Residue after Evaporation: 0.001% max. (ACS) Specific Gravity: ≤ 0.7962 at 15.56°C (USP) Substances Reducing Permanganate: Pass Test (ACS) Water: 0.2% max. (ACS) Benzene: 2ppm max. (USP, EP)
Ethanol, 200 proof (100%), USP, Decon™ Labs Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ethanol Name Note: 200 Proof (100%) CAS: 7732-18-5 CAS Min %: 99.9 CAS Max %: 100.0 Purity Grade: BP/EP/FCC/JP/NF/USP Synonym: Ethyl Alcohol
Ethanol, 95.5% (USP), Ricca Chemical
CAS: 64-17-5 CAS Min %: 98.0 CAS Max %: 100.0
Ethyl Alcohol Solution, 50% (v/v) Aqueous Solution, Ricca Chemical
Chemical Name or Material: Ethyl Alcohol CAS: 64-17-5 CAS: 7732-18-5 Physical Form: Liquid
Ethyl alcohol, Pure 200 Proof (Taxed), Meets USP testing specifications, for domestic sale in Ireland and United Kingdom only, Honeywell Riedel-de Haën™
Density: 0.790 - 0.791 g/cm3 Boiling Point: 78 to 79°C Linear Formula: CH3CH2OH Formula Weight: 46 Purity Grade: Pure CAS: 64-17-5 MDL Number: MFCD00003568 Flash Point: 12°C Chemical Name or Material: Ethyl alcohol, Pure (Taxed) Melting Point: −114°C CAS Max %: 100.0000% Beilstein: 1718733 Name Note: Meets USP testing specifications, for domestic sale in Germany, France, Ireland and United Kingdom Packaging: Glass Bottle Assay: >99.50% UN Number: UN1170
  spinner