Multicompendial IPA Isopropanol
2-Propanol (USP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: BP/EP/FCC/JP/NF/USP Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol, Isopropyl Alcohol
2-Propanol (USP/EP/BP/JP), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Purity Grade: BP/EP/FCC/JP/NF/USP Assay: ≥99.0% (USP) Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674 Synonym: Isopropanol, Isopropyl Alcohol
Isopropyl Alcohol USP, Macron Fine Chemicals™
Chemical Name or Material: Isopropyl Alcohol CAS: 67-63-0 Purity Grade: BP/EP/FCC/JP/NF/USP Molecular Formula: CH3CHOHCH3 Formula Weight: 60.1
Isopropyl Alcohol, Honeywell™ Riedel-de-Haën™
Vapor Pressure: 33mmHg (20°C), 44mmHg (25°C) Vapor Density: 2.1 (vs air) Density: 0.785g/mL at 25°C Explosion Limit: 0.02 Flash Point: 12°C (53.6°F) Ignition Point: 750°F Linear Formula: (CH3)2CHOH MDL Number: MFCD00011674 UN Number: UN1219 Formula Weight: 60.1g/mol Chemical Name or Material: 2-Propanol CAS: 67-63-0 Beilstein: 635639 Physical State: Liquid Color: Colorless Odor: Alcohol-like pH: Neutral Melting Point: −90°C Boiling Point: 77° - 83°C at 1,013 hPa Name Note: Meets USP testing specifications
Isopropyl Alcohol, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9%, Honeywell™
Density: 0.785 g/cm3 Boiling Point: 82.3°C Solubility: Miscible in Water at 20°C Formula Weight: 60.11 Purity Grade: for HPLC Water: 0.06% max. Solubility Information: Miscible in Water at 20°C CAS: 67-63-0 Synonym: 2-Propanol MDL Number: MFCD00011674 Flash Point: 12°C Chemical Name or Material: Isopropyl Alcohol CAS Max %: 100.0000% Beilstein: 635639 Name Note: ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9% Assay: >99.9% UN Number: UN1219
Isopropyl Alchohol, 99%, World/GMP, ACS/USP/FCC/EP/BP/JP Grade, PHARMCO-AAPER
Chemical Name or Material: Isopropyl Alcohol 99% CAS: 67-63-0 Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: IPA 99%, Isopropanol, 2-Propanol
2-Propanol, puriss., ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Vapor Pressure: 33mmHg (20°C), 44mmHg (25°C) Vapor Density: 2.1 (vs air) Density: 0.785g/mL at 25°C Explosion Limit: 0.02 Flash Point: 12°C (53.6°F) Ignition Point: 750°F Linear Formula: (CH3)2CHOH Name Note: Meets analytical specification of Ph. Eur.BP, USP MDL Number: MFCD00011674 UN Number: UN1219 Assay: ≥99.5% (GC) Formula Weight: 60.1g/mol Purity Grade: puriss. Chemical Name or Material: 2-Propanol CAS: 67-63-0 Beilstein: 635639 Boiling Point: 82°C Melting Point: −85°C Physical State: Liquid Color: Colorless
Isopropanol, 10% v/v (Certified), Fisher Chemical
Chemical Name or Material: Isopropanol, 10% v/v Name Note: Made from water and isopropyl alcohol CAS: 67-63-0 CAS: 7732-18-5 Purity Grade: Certified Grade Molecular Formula: C3H8O Linear Formula: (CH3)2CHOH Formula Weight: 60.1 Synonym: 10% 2-Propanol, IPA
Isopropyl Alcohol, 70%, GMP/USP/NF Grade, PHARMCO-AAPER
Chemical Name or Material: Isopropyl Alcohol 70% CAS: 67-63-0 CAS: 7732-18-5 Physical Form: Liquid Assay Percent Range: 70% Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: IPA 70%, Isopropanol, 2-Propanol
  spinner