Application Specific n- Hexane
n-Hexane, Environmental Grade, >95%, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >95% Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
n-Hexane, 95%, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane CAS: 110-54-3 CAS: 107-83-5 CAS: 96-14-0 Purity Grade: OPTIMA™ Purity Grade Notes: Also meets ACS specifications Assay Percent Range: 95% Molecular Formula: C6H14 Formula Weight: 86.17 MDL Number: MFCD00009520
n-Hexane, 97%, for electronic use, residue free, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: Residue Free CAS: 110-54-3 CAS Min %: 96.0 CAS Max %: 100.0 Purity Grade: For electronic use (MOS) Assay: 97% Assay Percent Range: 97% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, Spectrophotometric Grade, >95%, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >95% Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
n-hexane, 95+%, for HPLC, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: For HPLC CAS: 110-54-3 CAS Min %: 95.0 CAS Max %: 100.0 Purity Grade: For HPLC Assay: ≥95% Assay Percent Range: 95+% Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Synonym: Hex
n-Hexane, for residue and pesticides anal., ca. 95% n-Hexane, ACROS Organics™
Chemical Name or Material: n-Hexane Name Note: ECD Tested, for Pesticide Analysis, Residue <0.0003% CAS: 110-54-3 Molecular Formula: C6H14 Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520 Synonym: Hex
n-Hexane, Alfa Aesar™
Chemical Name or Material: n-Hexane CAS: 110-54-3 Physical Form: Liquid, Distilled in Glass Assay Percent Range: 95% min Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 MDL Number: MFCD00009520
Hexane, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.9% (>60% n-Hexane), Honeywell™
CAS Max %: <40.0000% Density: 0.659 - 0.673 g/cm3 Boiling Point: 68.7°C Solubility: 1.4% in Water at 20°C Formula Weight: 86.18 CAS Min %: >60.0000% Purity Grade: for HPLC Water: 0.01% max. Absorbance: 0.05 AU max. at 225 nm Solubility Information: 1.4% in Water at 20°C CAS: 110-54-3 Flash Point: −26°C Chemical Name or Material: Hexane CAS Max %: >60.00% Beilstein: 1730733 Name Note: ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.9% (>60% n-Hexane) Assay: >99.9% UN Number: UN1208
Hexane CHROMASOLV™, for HPLC, ≥97.0% (GC), Honeywell Riedel-de Haën™
Beilstein: 1730733 CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Assay: ≥97.0% (GC) Boiling Point: 69°C Formula Weight: 86.18g/mol Melting Point: −95°C Purity Grade: CHROMASOLV™ Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 Molecular Formula: C6H14 Name Note: for HPLC Refractive Index: n20/D 1.375 Synonym: n-Hexane Vapor Density: ˜3 (vs. air) Vapor Pressure: 256mmHg (37.7°C)
n-Hexane For spectroscopy Uvasol™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
n-Hexane For gas chromatography ECD and FID SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
n-Hexane, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Hexane Name Note: OmniPure™ CAS: 110-54-3 Assay Percent Range: 95% Linear Formula: CH3(CH2)4CH3 Formula Weight: 86.18
n-Hexane For gas chromatography MS SupraSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Hexane CAS: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 UN Number: 1208
Hexane, For pesticide residue analysis, Honeywell Riedel-de Haën™
Beilstein: 1730733 CAS: 110-54-3 Chemical Name or Material: Hexane Density: 0.659g/mL at 25°C Formula Weight: 86.18g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.077 Flash Point: −26°C (−14.8°F) Ignition Point: 453°F Linear Formula: CH3(CH2)4CH3 MDL Number: MFCD00009520 UN Number: UN1208 Vapor Density: 3 (vs air) Vapor Pressure: 5.2 psi (37.7°C), 256 mmHg (37.7°C), 132 mmHg (20°C)
Alfa Aesar™ n-Hexane, Spectrophotometric Grade, 95+%, packaged under Argon in resealable ChemSeal™ bottles
Chemical Name or Material: n-Hexane CAS: 110-54-3 Purity Grade: Spectrophotometric grade Formula Weight: 86.18 MDL Number: MFCD00009520 UN Number: UN1208 Beilstein: 1730733 Merck Index: 14,4694
Hexane, mixture of isomers, Puriss., ≥85% isomers basis (GC), Honeywell
Density: 0.672 g/mL at 25 °C(lit.) Flash Point: −26°C MDL Number: MFCD00004599 UN Number: UN1208 Assay: >85% Purity Grade: Puriss. Chemical Name or Material: Hexane, mixture of isomers Boiling Point: 48 to 65°C Melting Point: −95°C(lit.)
  spinner