1-Propanol
2-Propanol, anhydrous, >99.5%, Alfa Aesar™
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674
1-Propanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Certified Grade Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.09 MDL Number: MFCD00002941 Synonym: n-Propyl Alcohol
1-Propanol, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
1-Propanol, 99.5%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol Name Note: Extra Dry CAS: 71-23-8 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
1-Propanol, ACS, >99.5%, Alfa Aesar
Chemical Name or Material: 1-Propanol Name Note: In ACS analysis CAS: 71-23-8 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, anhydrous, 99.9%, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, 99+%, Extra Pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol CAS: 71-23-8 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
Alfa Aesar™ 1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, Acros Organics™
Density: 0.8000g/mL Health Hazard 3: GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Health Hazard 2: GHS H Statement May cause drowsiness or dizziness. Causes serious eye damage. Highly flammable liquid and vapor. Health Hazard 1: Danger Linear Formula: CH3CH2CH2OH Melting Point: −127.0°C Water: 0.2% max. Viscosity: 2.2 mPa.s (20°C) Formula Weight: 60.1 Solubility Information: Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents Purity Grade: ACS Grade Refractive Index: 1.3840 to 1.3860 Flash Point: 15°C Boiling Point: 97.0°C CAS: 71-23-8 Molecular Formula: C3H8O Assay: ≥99.5% (GC) 0.01% max. (Ethanol) (GC) 0.01% max. (Methanol) (GC) 0.05% max. (Isopropyl alcohol) (GC) Chemical Name or Material: 1-Propanol
1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol Products for Science Education
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Reagent Grade Assay Percent Range: >99% Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: N-Propyl Alcohol
n-Propyl Alcohol AR, Macron Fine Chemicals™
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Molecular Formula: CH3(CH2)2OH Formula Weight: 60.1
n-Propyl Alcohol ChromAR, Macron Fine Chemicals™
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Molecular Formula: CH3(CH2)2OH Formula Weight: 60.1
n-Propanol (Peroxide-Free/Sequencing), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Propanol Name Note: Peroxide-free CAS: 71-23-8 Purity Grade: Sequencing Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: 1-Propanol
n-Propyl Alcohol, OmniSolv™, EMD Millipore
Chemical Name or Material: OmniSolv™ n-Propyl Alcohol CAS: 71-23-8
n-Propyl Alcohol, 99.5% min.,OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: 1-Propanol
n-Propyl Alcohol, Reagent grade, 98.0% min., EMD Millipore
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: 98.0%, min. Molecular Formula: CH3CH2CH2OH Linear Formula: CH3CH2CH2OH Formula Weight: 6.1 Synonym: 1-Propanol
n-Propyl Alcohol, WORLD/GMP, ACS/FCC/EP Grade, PHARMCO-AAPER
Chemical Name or Material: n- Propyl Alcohol CAS: 71-23-8 Physical Form: Liquid Molecular Formula: C3H8O Formula Weight: 60.1 Synonym: 1-Propanol
  spinner