Analytical 1- Propanol
Alfa Aesar™ 1-Propanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, 99+%, Extra Pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol CAS: 71-23-8 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
2-Propanol, anhydrous, >99.5%, Alfa Aesar™
Chemical Name or Material: 2-Propanol CAS: 67-63-0 Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: (CH3)2CHOH Formula Weight: 60.1 MDL Number: MFCD00011674
1-Propanol (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Certified Grade Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.09 MDL Number: MFCD00002941 Synonym: n-Propyl Alcohol
1-Propanol Products for Science Education
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Reagent Grade Assay Percent Range: >99% Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: N-Propyl Alcohol
1-Propanol, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
1-Propanol, anhydrous, 99.9%, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Physical Form: Liquid Assay Percent Range: 99.9% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, 99.5%, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: 1-Propanol Name Note: Extra Dry CAS: 71-23-8 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.5% (GC) Assay Percent Range: 99.5% Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941 Synonym: Propyl alcohol
1-Propanol, Acros Organics™
Density: 0.8000g/mL Linear Formula: CH3CH2CH2OH Melting Point: −127.0°C Water: 0.2% max. Viscosity: 2.2 mPa.s (20°C) Formula Weight: 60.1 Solubility Information: Solubility in water: soluble. Other solubilities: miscible with alcohol and ether, soluble in many organic solvents Purity Grade: ACS Grade Refractive Index: 1.3840 to 1.3860 Flash Point: 15°C Boiling Point: 97.0°C CAS: 71-23-8 Molecular Formula: C3H8O Assay: ≥99.5% (GC) 0.01% max. (Ethanol) (GC) 0.01% max. (Methanol) (GC) 0.05% max. (Isopropyl alcohol) (GC) Chemical Name or Material: 1-Propanol
1-Propanol, ACS, >99.5%, Alfa Aesar
Chemical Name or Material: 1-Propanol Name Note: In ACS analysis CAS: 71-23-8 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99.5% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
1-Propanol, BAKER ANALYZED™ Reagent, J.T.Baker™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: BAKER ANALYZED™ Reagent Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1
n-Propyl Alcohol AR, Macron Fine Chemicals™
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Molecular Formula: CH3(CH2)2OH Formula Weight: 60.1
1-Propanol, purum, ≥99% (GC), Honeywell™
Vapor Pressure: 10mmHg (147°C), 14.9mmHg (20°C) Vapor Density: 2.1 (vs air) Density: 0.804g/mL at 25°C Explosion Limit: 0.137 Flash Point: 22°C (71.6°F) Ignition Point: 700°F Linear Formula: CH3CH2CH2OH MDL Number: MFCD00002941 UN Number: UN1274 Assay: ≥99% (GC) Formula Weight: 60.1g/mol Purity Grade: purum Chemical Name or Material: 1-Propanol CAS: 71-23-8 Beilstein: 1098242 Melting Point: −127° Physical State: Liquid
1-Propanol, ACS Reagent, ≥99.5%, Honeywell™ Riedel-de Haën™
Physical State: Liquid Color: APHA: ≤10 Density: 0.804g/mL at 25°C Vapor Pressure: 14.9mmHg (20°C), 10mmHg (147°C) Vapor Density: 2.1 (vs air) Flash Point: 22°C (71.6°F) Explosion Limit: 0.137 Ignition Point: 700°F Melting Point: −127°C Boiling Point: 97°C Refractive Index: n20/D 1.384
1-Propanol, puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell™ Riedel-de Haën™
Color: Colorless Odor: Alcohol-like Density: 0.804g/mL at 25°C Solubility: Soluble in water and most organic solvents Vapor Pressure: 14.9mmHg (20°C), 10mmHg (147°C) Vapor Density: 2.1 (vs air) Density: 0.804g/mL at 25°C Flash Point: 22°C (71.6°F) Ignition Point: 700°F Refractive Index: n20/D 1.384 Melting Point: −127°C Boiling Point: 97°C
  spinner