Application Specific 1- Propanol
1-Propanol, Spectrophotometric Grade, 99%, Alfa Aesar™
Chemical Name or Material: 1-Propanol CAS: 71-23-8 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 MDL Number: MFCD00002941
n-Propyl Alcohol ChromAR, Macron Fine Chemicals™
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Molecular Formula: CH3(CH2)2OH Formula Weight: 60.1
n-Propanol (Peroxide-Free/Sequencing), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Propanol Name Note: Peroxide-free CAS: 71-23-8 Purity Grade: Sequencing Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H8O Linear Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: 1-Propanol
n-Propyl Alcohol, 99.5% min.,OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Physical Form: Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3CH2CH2OH Formula Weight: 60.1 Synonym: 1-Propanol
n-Propyl Alcohol, OmniSolv™, EMD Millipore
Chemical Name or Material: OmniSolv™ n-Propyl Alcohol CAS: 71-23-8
n-Propyl Alcohol, Reagent grade, 98.0% min., EMD Millipore
Chemical Name or Material: n-Propyl Alcohol CAS: 71-23-8 Purity Grade: Reagent Grade Physical Form: Liquid Assay Percent Range: 98.0%, min. Molecular Formula: CH3CH2CH2OH Linear Formula: CH3CH2CH2OH Formula Weight: 6.1 Synonym: 1-Propanol
  spinner