Cyclohexane
Cyclohexane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥99.0% Assay Percent Range: 99.0% min by GC Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Certified ACS Reagent Grade Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, for residue analysis, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: For residue analysis Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, HPLC Grade, 99% min.; Alfa Aesar™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: C6H12 Formula Weight: 84.16 MDL Number: MFCD00003814
Cyclohexane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Spectranalyzed™ Purity Grade Notes: Also meets ACS specifications Assay: ≥99.0% Assay Percent Range: ≥99.0 % Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, 99+%, pure, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, 99.5%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, 99.8%, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: For HPLC Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane (Reagent), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Reagent Grade Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane Name Note: 99+% CAS: 110-82-7 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane (Pesticide), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Pesticide Grade Molecular Formula: C6H12 Linear Formula: CH2(CH2)4CH2 Formula Weight: 84.16 MDL Number: MFCD00003814 Synonym: Hexahydrobenzene
Cyclohexane, for HPLC, ACROS Organics™
Chemical Name or Material: Cyclohexane Name Note: 99.0% CAS: 110-82-7 Purity Grade: For HPLC Assay: ≥99.0% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, >99%, ACS, Alfa Aesar™
Chemical Name or Material: Cyclohexane Name Note: For ACS analysis CAS: 110-82-7 Purity Grade: ACS Grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C6H12 Formula Weight: 84.16 MDL Number: MFCD00003814
Cyclohexane, extra pure, ACROS Organics™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Pure Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15
Cyclohexane, ≥99.8%, Laboratory Reagent, Honeywell™
Vapor Pressure: 168.8mmHg (37.7°C), 77mmHg (20°C) Vapor Density: 2.9 (vs air) Density: 0.779g/mL at 25°C Explosion Limit: 0.09 Ignition Point: 500°F MDL Number: MFCD00003814 UN Number: UN1145 Assay: ≥99.8% Formula Weight: 84.16g/mol Chemical Name or Material: Cyclohexane CAS: 110-82-7 Beilstein: 1900225 Physical State: Liquid Color: Colorless Melting Point: 6.4°C Boiling Point: 81°C at 1013 hPa Odor: Characteristic
Alfa Aesar™ Cyclohexane, Environmental Grade, 99.7+%
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Environmental Grade Formula Weight: 84.16 MDL Number: MFCD00003814 UN Number: UN1145 Beilstein: 1900225 Merck Index: 14,2723
Cyclohexane, 99+%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: For spectroscopy ACS Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Chemical Name or Material: Cyclohexane Name Note: Extra Dry CAS: 110-82-7 Purity Grade: Extra Dry over Molecular Sieve Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC), Honeywell Riedel-de Haën™
Vapor Pressure: 168.8mmHg (37.7°C), 77mmHg (20°C) Vapor Density: 2.9 (vs air) Density: 0.779g/mL at 25°C Color: APHA: ≤10 Explosion Limit: 0.09 Flash Point: −18°C (−.4°F) Ignition Point: 500°F Refractive Index: n20/D 1.4250 to 1.4280 Boiling Point: 79° to 81°C Melting Point: 4 to 7°C Residue after Evaporation: 0 to 0.001% Impurities: ≤0.05% cyclohexene (C6H10)
Alfa Aesar™ Cyclohexane, 99+%
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16 MDL Number: MFCD00003814 UN Number: UN1145 Beilstein: 1900225 Merck Index: 14,2723
Cyclohexane, 99.0%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99.0% Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, for residue analysis, suitable for residue and pesticides analysis, ACROS Organics™
Chemical Name or Material: Cyclohexane Name Note: ECD Tested, For Pesticide Analysis CAS: 110-82-7 Purity Grade: For pesticide residue analysis Molecular Formula: C6H12 Linear Formula: C6H12 Formula Weight: 84.15 MDL Number: MFCD00003814
Cyclohexane, HPLC Grade, J.T.Baker™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: HPLC Grade Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane, ULTRA RESI-ANALYZED™, J.T.Baker™
Chemical Name or Material: Cyclohexane CAS: 110-82-7 Purity Grade: ULTRA RESI-ANALYZED™ Molecular Formula: C6H12 Formula Weight: 84.16
Cyclohexane, AR ACS), For ACS analysis, Macron Fine Chemicals™
Chemical Name or Material: Cyclohexane Name Note: ACS CAS: 110-82-7 Purity Grade: ACS Reagent Grade Assay Percent Range: 99.0 % min. Molecular Formula: C6H12 Formula Weight: 84.16
  spinner