Formamide
Formamide (Certified ACS), Fisher Chemical
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Certified ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941 Synonym: Methanamide
Alfa Aesar™ Formamide, ACS, 99.5+%
Chemical Name or Material: Formamide CAS: 75-12-7 Name Note: For ACS analysis Purity Grade: ACS Grade Molecular Formula: HCONH2 Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941 Beilstein: 505995 Merck Index: 14,4237
Formamide, 99%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: DNase, RNase and Protease Free CAS: 75-12-7 Purity Grade: For molecular biology Assay: 99% Assay Percent Range: 99% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide (Molecular Biology), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Physical Form: Liquid Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: CH3NO Linear Formula: H2NCH:O Formula Weight: 45.04
Formamide, 99+%, for spectroscopy, ACROS Organics™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For spectroscopy Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, 99.5+%, extra pure, ACROS Organics™
Chemical Name or Material: Formamide Name Note: 99.5+% CAS: 75-12-7 Purity Grade: Extra Pure Assay: ≥99.5% Assay Percent Range: 99.5+% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide (≥99.5%), Fisher BioReagents Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Biotech Grade Physical Form: Liquid Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: CH3NO Linear Formula: H2NCH:O Formula Weight: 45.04
Formamide, 99.5%, for analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: For analysis Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, 99.6%, ACS reagent, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: ACS Reagent Grade Assay: ≥99.5% Assay Percent Range: 99.6% Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide OR, J.T.Baker™
Chemical Name or Material: Formamide CAS: 75-12-7 Molecular Formula: HCONH2 Formula Weight: 45.04
Formamide, Deionized, OmniPur™, Calbiochem™, EMD Millipore
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Molecular Formula: CH3NO Formula Weight: 45 Synonym: Carbamaldehyde
Formamide, Ultrapure Bioreagent, J.T.Baker™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Ultrapure Bioreagent Molecular Formula: HCONH2 Formula Weight: 45.04
Formamide, 99.5% min., ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Formamide Name Note: For ACS analysis CAS: 75-12-7 Purity Grade: ACS/EP/Reagent Grade Physical Form: Slightly Viscous Liquid Assay Percent Range: 99.5% min. Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.4 Synonym: Formic acid amide, Methane amide
Alfa Aesar™ Formamide, 99%
Chemical Name or Material: Formamide CAS: 75-12-7 Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941 Beilstein: 505995 Merck Index: 14,4237
Alfa Aesar™ Formamide, Molecular Biology Grade
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Formula Weight: 45.04 MDL Number: MFCD00007941 Beilstein: 505995 Merck Index: 14,4237
Formamide, puriss. p.a., ACS Reagent, ≥99.5% (GC/T), Honeywell™ Riedel-de Haën™
Physical State: Liquid Color: Colorless Odor: Weak pH: 4 to 5 at 20°C Density: 1.132g/mL at 20°C, 1.134g/mL at 25°C Vapor Density: 1.55 (vs air) Vapor Pressure: 30mmHg (129°C), 0.08mmHg (20°C) Flash Point: 175°C (347°F) Ignition Point: 932°F Melting Point: 1 to 3°C Boiling Point: 210°C
Formamide, Molecular Biology Grade Calbiochem™, EMD Millipore
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Molecular Biology Grade Molecular Formula: CH3NO Formula Weight: 45 Synonym: Carbamaldehyde, Methanamide
Alfa Aesar™ Formamide, Spectrophotometric Grade, 99+%
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Spectrophotometric grade Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941 Beilstein: 505995 Merck Index: 14,4237
Alfa Aesar™ Formamide, Ultra Pure, 99.5% min
Chemical Name or Material: Formamide CAS: 75-12-7 Name Note: Ultra Pure Formula Weight: 45.04 MDL Number: MFCD00007941 Beilstein: 505995
Formamide, ≥99%, Ultrapure, Affymetrix/USB™
Chemical Name or Material: Formamide CAS: 75-12-7 Assay: ≥99% Formula Weight: 45.04 Physical Form: Liquid Linear Formula: HCONH2 MDL Number: MFCD00007941 Molecular Formula: CH3NO Synonym: Methanamide Boiling Point: 210°C Melting Point: 2°C Flash Point: 154°C (309°F) Specific Gravity: 1.134 Purity Grade: Ultrapure
Formamide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: HCONH2 Formula Weight: 45.04
Formamide, ≥99.0% (GC), Reagent Grade, Honeywell™
Vapor Pressure: 0.08mmHg (20°C), 30mmHg (129°C) Vapor Density: 1.55 (vs air) Density: 1.134g/mL at 25°C Flash Point: 175°C (347°F) Ignition Point: 932°F Linear Formula: HCONH2 MDL Number: MFCD00007941 UN Number: NONH for all modes of transport Assay: ≥99.0% (GC) Formula Weight: 45.04g/mol Chemical Name or Material: Formamide CAS: 75-12-7 Beilstein: 505995 Physical State: Liquid Color: Colorless Odor: Weak pH: 4.0 - 5.0 at 20°C Melting Point: 1 - 3°C Boiling Point: 210°C at 1,013 hPa Solubility in Water: Completely miscible
Formamide, Spectrophotometric Grade, ≥99%, Honeywell Riedel-de Haën™
Vapor Pressure: 0.08mmHg (20°C), 30mmHg (129°C) Vapor Density: 1.55 (vs air) Density: 1.134g/mL at 25°C Flash Point: 175°C (347°F) Ignition Point: 932°F Linear Formula: HCONH2 MDL Number: MFCD00007941 UN Number: NONH for all modes of transport Assay: ≥99% Purity Grade: Spectrophotometric grade Chemical Name or Material: Formamide CAS: 75-12-7 Beilstein: 505995 Boiling Point: 210°C Melting Point: 1° to 3°C Refractive Index: 1.447
Formamide, Polarographic and Nonaqueous, Titration Grade, ACROS Organics™
Chemical Name or Material: Formamide CAS: 75-12-7 Purity Grade: Analytical Grade Molecular Formula: CH3NO Linear Formula: HCONH2 Formula Weight: 45.04 MDL Number: MFCD00007941
Formamide, BAKER™, J.T.Baker™
Chemical Name or Material: Formamide CAS: 75-12-7 Molecular Formula: HCONH2 Formula Weight: 45.04
  spinner