n-Heptane
Heptane (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Certified Grade Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane (Technical), Fisher Chemical
Chemical Name or Material: Heptane CAS: 142-82-5 Purity Grade: Technical Grade Molecular Formula: C7H16 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Extra Pure Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Over Molecular Sieves CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, HPLC Grade, greater than99%, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
n-Heptane, 99%, for molecular biology, sequenation grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane Name Note: 99% CAS: 142-82-5 Purity Grade: For molecular biology Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C7 H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane Products for Science Education
Chemical Name or Material: Heptane CAS: 142-82-5 Linear Formula: C7H16 Formula Weight: 100.21 Synonym: N-Heptane
n-Heptane, 99+%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Water <30ppm CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, Environmental Grade, 96+%, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >96% Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
n-Heptane, Spectrophotometric Grade, greater-than99% n-Heptane 96% min, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
n-Heptane, 96%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Extra Dry Assay: ≥96% Assay Percent Range: 96% Molecular Formula: C7 H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99%, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: OPTIMA™ Assay Percent Range: 99.0% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Tested for Trace Analysis of Polyaromatic Hydrocarbons CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-heptane, for HPLC, ∽ 95% n-heptane, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: For HPLC Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥96% Assay Percent Range: 96% min by GC Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Spectranalyzed™ Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane Name Note: ECD Tested, For Pesticide Analysis, Sulfur Low CAS: 142-82-5 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, for HPLC, (approx. 99% n-Heptane), ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: For HPLC Assay: ≥99% (GC) Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Heptane CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane (Peroxide-free/Sequencing), Fisher BioReagents
Chemical Name or Material: Heptane Name Note: Peroxide-free CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Sequencing Grade Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Heptane CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane, CHROMASOLV™, ≥99%, LC-MS, Honeywell Riedel-de Haën™
CAS: 142-82-5 Chemical Name or Material: Heptane Density: 0.684g/mL at 25°C Assay: ≥99% Formula Weight: 100.2 Explosion Limit: 0.07 Flash Point: −4°C (24.8°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331-P501 Ignition Point: 433°F Linear Formula: CH3(CH2)5CH3 MDL Number: MFCD00009544 RTECS Number: 205-563-8 UN Number: UN1206 Vapor Density: 3.5 (vs air) Vapor Pressure: 83 mmHg (37.7°C), 40 mmHg (20°C)
Heptane, CHROMASOLV™, ≥99% (GC), Honeywell Riedel-de Haën™
CAS: 142-82-5 Chemical Name or Material: Heptane Density: 0.684g/mL at 25°C Assay: ≥99% (GC) Formula Weight: 100.2 Explosion Limit: 0.07 Flash Point: −4°C (24.8°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331-P501 Ignition Point: 433°F Linear Formula: CH3(CH2)5CH3 MDL Number: MFCD00009544 RTECS Number: 205-563-8 UN Number: UN1206 Vapor Density: 3.5 (vs air) Vapor Pressure: 83 mmHg (37.7°C), 40 mmHg (20°C)
n-Heptane, >99%, MP Biomedicals™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C7H16 Formula Weight: 100.21 Synonym: Dipropyl methane
Heptane, 99.0% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Synonym: n-Heptane
n-Heptane, 99%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Assay Percent Range: 0.99
n-Heptane For spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: n-Heptane CAS: 142-82-5 Molecular Formula: C7H16 Formula Weight: 100.2 UN Number: 1206
  spinner