Analytical n- Heptane
n-Heptane, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Extra Pure Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, 99+%, pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Pure Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 96%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Extra Dry Assay: ≥96% Assay Percent Range: 96% Molecular Formula: C7 H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, Extra Dry, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Water <30ppm CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane (Technical), Fisher Chemical
Chemical Name or Material: Heptane CAS: 142-82-5 Purity Grade: Technical Grade Molecular Formula: C7H16 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane (Certified), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Certified Grade Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99.5%, for analysis, ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For analysis Assay: ≥99.4% Assay Percent Range: 99.5% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane Products for Science Education
Chemical Name or Material: Heptane CAS: 142-82-5 Linear Formula: C7H16 Formula Weight: 100.21 Synonym: N-Heptane
n-heptane, for HPLC, ∽ 95% n-heptane, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: For HPLC Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Over Molecular Sieves CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: ≥99% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Heptane CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane (Peroxide-free/Sequencing), Fisher BioReagents
Chemical Name or Material: Heptane Name Note: Peroxide-free CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Sequencing Grade Assay: ≥99% Assay Percent Range: ≥99 % Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, >99%, MP Biomedicals™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: C7H16 Formula Weight: 100.21 Synonym: Dipropyl methane
n-Heptane, 99%, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Assay Percent Range: 0.99
Heptane, 99.0% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 Physical Form: Liquid Assay Percent Range: 99.0% min. Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Synonym: n-Heptane
Heptane, EMD Millipore™
CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 Chemical Name or Material: Heptane
n-Heptane, 99%, GMP/High Purity Grade, PHARMCO-AAPER
Chemical Name or Material: n-Heptane 99% CAS: 142-82-5 Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: C7H16 Formula Weight: 100.2 Synonym: Heptane
n-Heptane, >95% min., Ultra Scientific
Chemical Name or Material: n-Heptane CAS: 000142-82-5 CAS Max %: 100.0 Assay Percent Range: >95% min.
Heptane, GMP/ACS Grade, PHARMCO-AAPER
Chemical Name or Material: Heptane CAS: 142-82-5 Physical Form: Liquid Molecular Formula: C7H16 Formula Weight: 100.2 Synonym: Heptane solution
n-Heptane AR, Macron™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.2
  spinner