Application Specific n- Heptane
Heptane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: Spectranalyzed™ Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, ACROS Organics™
Chemical Name or Material: n-Heptane Name Note: Tested for Trace Analysis of Polyaromatic Hydrocarbons CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, Environmental Grade, 96+%, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >96% Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
n-Heptane, 99+%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane Name Note: ECD Tested, For Pesticide Analysis, Sulfur Low CAS: 142-82-5 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥99.0% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99+%, for spectroscopy, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99% (GC) Assay Percent Range: 99+% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
Heptane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Heptane CAS: 142-82-5 CAS: 28729-52-4 CAS: 26635-64-3 CAS: 108-87-2 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: ≥96% Assay Percent Range: 96% min by GC Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, for HPLC, (approx. 99% n-Heptane), ACROS Organics™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: For HPLC Assay: ≥99% (GC) Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, 99%, Optima™, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: OPTIMA™ Assay Percent Range: 99.0% Molecular Formula: C7H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2
n-Heptane, HPLC Grade, greater than99%, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
n-Heptane, 99%, for molecular biology, sequenation grade, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Heptane Name Note: 99% CAS: 142-82-5 Purity Grade: For molecular biology Assay: ≥98.5% Assay Percent Range: 99% Molecular Formula: C7 H16 Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.2 MDL Number: MFCD00009544
n-Heptane, Spectrophotometric Grade, greater-than99% n-Heptane 96% min, Alfa Aesar™
Chemical Name or Material: n-Heptane CAS: 142-82-5 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.21 MDL Number: MFCD00009544
Heptane ChromAR, Macron Fine Chemicals™
Chemical Name or Material: Heptane CAS: 142-82-5 Molecular Formula: CH3(CH2)5CH3 Formula Weight: 100.2
Heptane, CHROMASOLV™, ≥99%, LC-MS, Honeywell Riedel-de Haën™
CAS: 142-82-5 Chemical Name or Material: Heptane Density: 0.684g/mL at 25°C Assay: ≥99% Formula Weight: 100.2 Explosion Limit: 0.07 Flash Point: −4°C (24.8°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331-P501 Ignition Point: 433°F Linear Formula: CH3(CH2)5CH3 MDL Number: MFCD00009544 RTECS Number: 205-563-8 UN Number: UN1206 Vapor Density: 3.5 (vs air) Vapor Pressure: 83 mmHg (37.7°C), 40 mmHg (20°C)
Heptane, CHROMASOLV™, ≥99% (GC), Honeywell Riedel-de Haën™
CAS: 142-82-5 Chemical Name or Material: Heptane Density: 0.684g/mL at 25°C Assay: ≥99% (GC) Formula Weight: 100.2 Explosion Limit: 0.07 Flash Point: −4°C (24.8°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331-P501 Ignition Point: 433°F Linear Formula: CH3(CH2)5CH3 MDL Number: MFCD00009544 RTECS Number: 205-563-8 UN Number: UN1206 Vapor Density: 3.5 (vs air) Vapor Pressure: 83 mmHg (37.7°C), 40 mmHg (20°C)
n-Heptane For spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: n-Heptane CAS: 142-82-5 Molecular Formula: C7H16 Formula Weight: 100.2 UN Number: 1206
Heptane, OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: OmniSolv™ Heptane
Heptane, EMD Millipore
Chemical Name or Material: Heptane Linear Formula: CH3(CH2)5CH3 Formula Weight: 100.21
Heptane OmniSolv™, EMD Millipore
Chemical Name or Material: Heptane CAS: 142-82-5 Molecular Formula: C7H16 Formula Weight: 100.2 UN Number: 1206 Synonym: n-Heptane
Heptane CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™
Beilstein: 1730763 CAS: 142-82-5 Chemical Name or Material: Heptane Density: 0.684g/mL at 25°C Assay: ≥99% Boiling Point: 98°C Color: APHA: ≤10 Formula Weight: 100.2g/mol Melting Point: −91°C Purity Grade: CHROMASOLV™ EINECS Number: 205-563-8 Health Hazard 2: P210-P273-P301 + P310-P304 + P340 + P312-P331-P391 Health Hazard 3: H225-H304-H315-H336-H410 Linear Formula: CH3(CH2)5CH3 MDL Number: MFCD00009544 Molecular Formula: C7H16 Name Note: for HPLC Purity Grade Notes: filtration through 0.2µm Refractive Index: n20/D 1.387 RTECS Number: MI7700000 Vapor Density: 3.5 (vs. air) Vapor Pressure: 83mmHg (37.7°C)
  spinner