N N-Dimethylformamide
N,N-Dimethylformamide (Certified ACS), Fisher Chemical
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Certified ACS Reagent Grade Physical Form: Liquid Assay: ≥99.8% Assay Percent Range: ≥99.8 % Molecular Formula: C3H7NO MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.5%, Acros Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.4 CAS Max %: 100.0 Purity Grade: For HPLC Assay: 99.5% Assay Percent Range: 99.5% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 MDL Number: MFCD00003284 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N Synonym: DMF PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Extra Dry Assay: ≥99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: Sequencing Grade Assay: ≥99.5% Assay Percent Range: ≥99.5 % Molecular Formula: C3H7NO Linear Formula: HCONCH3CH3 Synonym: DMF
DMF, Dimethylformamide, GC Headspace Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N Synonym: N,N-Dimethylmethanamide, N,N-Dimethylformamide PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, 99.8%, for peptide synthesis, ACROS Organics™
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For peptide synthesis Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide Name Note: water <50 ppm CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry over Molecular Sieve Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, HPLC Grade, greater-than99.7%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C3H7NO MDL Number: MFCD00003284
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: Extra Dry Purity Grade Notes: Water <50ppm Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%, Honeywell Burdick & Jackson
Density: 0.949 g/cm3 Boiling Point: 153°C Solubility: Miscible in Water at 20°C Purity Grade: for HPLC Water: 0.03% max. Solubility Information: Miscible in Water at 20°C CAS: 68-12-2 Flash Point: 58°C Chemical Name or Material: N,N-Dimethylformamide CAS Max %: 100.0000% Beilstein: 605365 Name Note: B&J Brand™, for HPLC, GC and spectrophotometry, >99.9% Packaging: Glass Bottle Assay: >99.9% UN Number: UN2265
N,N-Dimethylformamide, 99.9% (for HPLC and Spectrophotometry), OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, ACS, 99.8+%, Alfa Aesar™
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 Purity Grade: ACS Grade Molecular Formula: C3H7NO MDL Number: MFCD00003284 UN Number: UN2265 Beilstein: 605365 Merck Index: 14,3243
N,N-Dimethylformamide, anhydrous, 99.8%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Physical Form: Liquid Assay Percent Range: 99.8% Molecular Formula: C3H7NO MDL Number: MFCD00003284
N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™ package of 4x25ML bottles, ACROS Organics™
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 MDL Number: MFCD00003284 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N Synonym: DMF PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, DriSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N Synonym: Formic Acid Dimethylamide PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Beilstein: 605365 CAS: 68-12-2 Chemical Name or Material: N,N-Dimethylformamide Density: 0.944g/mL Assay: ≥99.9% Boiling Point: 153°C Melting Point: −61°C Purity Grade: CHROMASOLV™ Plus Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Molecular Formula: C3H7NO Name Note: for HPLC Purity Grade Notes: glass distillation Refractive Index: n20/D 1.430 Synonym: DMF; NSC 5356 Vapor Density: 2.5 (vs. air) Vapor Pressure: 2.7mmHg (20 °C)
N,N-Dimethylformamide, Anhydrous, 99.8% min., DriSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N Synonym: Formic Acid Dimethylamide PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, HPLC grade, >99.7%, Alfa Aesar™
Chemical Name or Material: N, N-Dimethylformamide CAS: 68-12-2 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: >99.7% Molecular Formula: C3H7NO MDL Number: MFCD00003284
Dimethylformamide, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
Chemical Name or Material: Dimethylformamide CAS: 68-12-2 Purity Grade: Baker Analyzed™ ACS Reagent Molecular Formula: C3H7NO
N,N-Dimethylformamide, 99.8+%, for spectroscopy ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide Name Note: 99.8% CAS: 68-12-2 CAS Min %: 99.8 CAS Max %: 100.0 Purity Grade: For spectroscopy ACS Assay: ≥99.8;% Assay Percent Range: 99.8+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 Molecular Formula: C3H7NO UN Number: 2265 Synonym: DMF, N,N-Dimethylmethanamide, Formic acid dimethylamide
N,N-Dimethylformamide, 99.8%, for molecular biology, DNAse, RNAse and Protease free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide Name Note: DNase, RNase, Protease Free CAS: 68-12-2 CAS Min %: 99.75 CAS Max %: 100.0 Purity Grade: For molecular biology Assay: 99.8% Assay Percent Range: 99.8% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylacetamide, CHROMASOLV™ Plus, for HPLC, ≥99.9%, Honeywell Riedel-de Haën™
Beilstein: 1737614 CAS: 127-19-5 Chemical Name or Material: N,N-Dimethylacetamide Density: 0.937g/mL at 25°C Assay: ≥99.9% Boiling Point: 164.5 to 166°C Melting Point: −20°C Purity Grade: CHROMASOLV™ Plus Linear Formula: CH3CON(CH3)2 MDL Number: MFCD00008686 Molecular Formula: C4H9NO Name Note: for HPLC Refractive Index: n20/D 1.437 Vapor Density: 3 (vs. air) Vapor Pressure: 2mmHg (25°C)
N,N-Dimethylformamide, 99+%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: N,N-Dimethylformamide Name Note: 99% CAS: 68-12-2 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 Synonym: DMF
N,N-Dimethylformamide, 99.8% (ACS Guaranteed Reagent), EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N PubChem CID: 6228 ChEBI: CHEBI:17741
N,N-Dimethylformamide, Anhydrous, septum-sealed, >99.9%, Honeywell Burdick & Jackson
Density: 0.949 g/cm3 Boiling Point: 153°C Solubility: Miscible in Water at 20°C Purity Grade: Septum-sealed Water: 50 ppm max. Solubility Information: Miscible in Water at 20°C CAS: 68-12-2 Physical Form: anhydrous Flash Point: 58°C Chemical Name or Material: N,N-Dimethylformamide CAS Max %: 100.0000% Beilstein: 605365 Name Note: Anhydrous, septum-sealed, for applications requiring low water content, >99.9% Packaging: Septum Sealed Bottle Assay: >99.9% UN Number: UN2265
N,N-Dimethylformamide, 99.8+%, ACS reagent, ACROS Organics™
Chemical Name or Material: N,N-Dimethylformamide CAS: 68-12-2 CAS Min %: 99.8 CAS Max %: 100.0 Purity Grade: ACS Reagent Grade Assay: ≥99.8;% Assay Percent Range: 99.8+% Molecular Formula: C3H7NO Linear Formula: HCON(CH3)2 Synonym: DMF
N,N-Dimethylformamide, Reagent Grade, ≥99%, Honeywell
Assay: >99.0% Beilstein: 605365 CAS: 68-12-2 Chemical Name or Material: N,N-Dimethylformamide Density: 0.944 g/mL(lit.) Explosion Limit: 0.152 Flash Point: 57.5°C Ignition Point: 833°F Linear Formula: HCON(CH3)2 MDL Number: MFCD00003284 UN Number: UN2265 Vapor Density: 2.5 (vs air) Vapor Pressure: 2.7 mmHg (20°C)
N,N-Dimethylformamide, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
CAS: 68-12-2 Molecular Formula: C3H7NO Molecular Weight (g/mol): 73.095 InChI Key: ZMXDDKWLCZADIW-UHFFFAOYSA-N PubChem CID: 6228 ChEBI: CHEBI:17741
  spinner