n-Pentane
n-Pentane, Spectrophotometric Grade, >99% min., Alfa Aesar™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, extra pure, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99.0% Assay Percent Range: 99+% Assay Percent Notes: Determine assay (GC) instead of accepting value from supplier's Certificate of Analysi Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 99% min., HPLC Grade, Alfa Aesar™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 99+%, Acros Organics™
Density: 0.6260g/mL Health Hazard 3: GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Health Hazard 2: GHS H Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking. Health Hazard 1: Danger Linear Formula: CH3(CH2)3CH3 Melting Point: −130.0°C Water: 0.005% max. Viscosity: 0.25 mPa.s (20°C) Formula Weight: 72.15 Solubility Information: Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents Purity Grade: Extra Dry over Molecular Sieve Refractive Index: 1.358 Flash Point: −49°C Boiling Point: 36.0°C CAS: 109-66-0 Molecular Formula: C5H12 Assay: ≥99.0% (GC) Chemical Name or Material: n-Pentane
n-Pentane, Environmental Grade, >98+%, Alfa Aesar™
Chemical Name or Material: n-Pentane Name Note: Environmental Grade CAS: 109-66-0 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >98% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: For HPLC Assay: ≥99.0% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 95%, technical grade, ACROS Organics™
Chemical Name or Material: n-Pentane CAS: 109-66-0 CAS Min %: 94.0 CAS Max %: 100.0 Assay: ≥94% Assay Percent Range: 95% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
n-Pentane, for analysis, ACROS Organics™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: For analysis Assay: ≥99.0% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: >99% Assay Percent Range: >99% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 98%
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498 UN Number: UN1265 Merck Index: 14,7116
n-Pentane, +99%, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane Name Note: Extra Dry CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Dry Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, extra pure, anhydrous, ACROS Organics™
Chemical Name or Material: n-Pentane Name Note: p.a. CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: Extra Pure Assay: ≥99% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, for spectroscopy, ACROS Organics™
Chemical Name or Material: n-Pentane Name Note: 99+% CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (Certified), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Certified Grade Assay: ≥98% Assay Percent Range: ≥98 % Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (Pesticide), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Pesticide Grade Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 99+%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane Name Note: ECD Tested, for Pesticide Analysis CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥98.5% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane Products for Science Education
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Reagent Grade Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3
Pentane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 CAS: 287-92-3 CAS: 78-78-4 Purity Grade: Spectranalyzed™ Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Pentane CAS: 109-66-0 Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15
Pentane, 98% min., OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Physical Form: Liquid Assay Percent Range: 98% min. Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
n-Pentane For spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 UN Number: 1265
Pentane, 98% min., EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Physical Form: Liquid Assay Percent Range: 98% Min. Molecular Formula: CH3(CH2)3CH3 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
n-Pentane For gas chromatography ECD and FID SupraSolv™, EMD Millipore
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 UN Number: 1265
Pentane Anhydrous, DriSolv™, EMD Millipore
Chemical Name or Material: Pentane Anhydrous CAS: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 UN Number: 1265 Synonym: n-Pentane
Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™
Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99.0% Boiling Point: 35to 36°C Formula Weight: 72.15g/mol Melting Point: −130°C Purity Grade: CHROMASOLV™ EINECS Number: 203-692-4 Health Hazard 2: P210-P261-P273-P301 + P310-P331 Health Hazard 3: H225-H304-H336-H411 Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 Molecular Formula: C5H12 Name Note: for HPLC Refractive Index: n20/D 1.358 RTECS Number: RZ9450000 Vapor Density: 2.48 (vs. air)
Pentane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
Additional Information: for pesticide residue analysis Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99% (GC) Formula Weight: 72.15g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.083 Flash Point: −49°C (−56.2°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331 Ignition Point: 500°F Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 RTECS Number: RZ9450000 UN Number: UN1265 Vapor Density: 2.48 (vs air)
Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Honeywell Riedel-de Haën™
Additional Information: for residue analysis (of high-volatile halogenated hydrocarbons) Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99%, ≥99% (GC) Formula Weight: 72.15g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.083 Flash Point: −49°C (−56.2°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331 Ignition Point: 500°F Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 RTECS Number: RZ9450000 UN Number: UN1265 Vapor Density: 2.48 (vs air)
Pentane Products for Science Education
Chemical Name or Material: Pentane CAS: 109-66-0 Assay Percent Range: 100% Molecular Formula: C5H12 Formula Weight: 72.15 Synonym: Amyl Hydride
  spinner