Application Specific n- Pentane
n-Pentane, Spectrophotometric Grade, >99% min., Alfa Aesar™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: Spectrophotometric grade Physical Form: Liquid Assay Percent Range: >99% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 99% min., HPLC Grade, Alfa Aesar™
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: HPLC Grade Physical Form: Liquid Assay Percent Range: 99% min. Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, Environmental Grade, >98+%, Alfa Aesar™
Chemical Name or Material: n-Pentane Name Note: Environmental Grade CAS: 109-66-0 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >98% Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, for HPLC, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane CAS: 109-66-0 Purity Grade: For HPLC Assay: ≥99.0% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (HPLC), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: HPLC Grade Purity Grade Notes: Mobile phase for HPLC applications Assay: >99% Assay Percent Range: >99% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, +99%, for spectroscopy, ACROS Organics™
Chemical Name or Material: n-Pentane Name Note: 99+% CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For spectroscopy Assay: ≥99.0% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (Pesticide), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 Purity Grade: Pesticide Grade Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
n-Pentane, 99+%, for residue analysis, ECD tested for pesticide analysis, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: n-Pentane Name Note: ECD Tested, for Pesticide Analysis CAS: 109-66-0 CAS Min %: 99.0 CAS Max %: 100.0 Purity Grade: For residue analysis Assay: ≥98.5% Assay Percent Range: 99+% Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane (Spectranalyzed™), Fisher Chemical
Chemical Name or Material: Pentane CAS: 109-66-0 CAS: 287-92-3 CAS: 78-78-4 Purity Grade: Spectranalyzed™ Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15 MDL Number: MFCD00009498
Pentane, 98% min., OmniSolv™, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Pentane CAS: 109-66-0 Physical Form: Liquid Assay Percent Range: 98% min. Molecular Formula: C5H12 Linear Formula: CH3(CH2)3CH3 Formula Weight: 72.15
n-Pentane For spectroscopy Uvasol™, EMD Millipore
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 UN Number: 1265
n-Pentane For gas chromatography ECD and FID SupraSolv™, EMD Millipore
Chemical Name or Material: n-Pentane CAS: 109-66-0 Molecular Formula: C5H12 Formula Weight: 72.15 UN Number: 1265
Pentane CHROMASOLV™, for HPLC, ≥99.0%, Honeywell Riedel-de Haën™
Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99.0% Boiling Point: 35to 36°C Formula Weight: 72.15g/mol Melting Point: −130°C Purity Grade: CHROMASOLV™ EINECS Number: 203-692-4 Health Hazard 2: P210-P261-P273-P301 + P310-P331 Health Hazard 3: H225-H304-H336-H411 Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 Molecular Formula: C5H12 Name Note: for HPLC Refractive Index: n20/D 1.358 RTECS Number: RZ9450000 Vapor Density: 2.48 (vs. air)
Pentane, CHROMASOLV™, for pesticide residue analysis, Honeywell Riedel-de Haën™
Additional Information: for pesticide residue analysis Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99% (GC) Formula Weight: 72.15g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.083 Flash Point: −49°C (−56.2°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331 Ignition Point: 500°F Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 RTECS Number: RZ9450000 UN Number: UN1265 Vapor Density: 2.48 (vs air)
Pentane, CHROMASOLV™, for residue analysis (of high-volatile halogenated hydrocarbons), Honeywell Riedel-de Haën™
Additional Information: for residue analysis (of high-volatile halogenated hydrocarbons) Beilstein: 969132 CAS: 109-66-0 Chemical Name or Material: Pentane Density: 0.626g/mL at 25°C Assay: ≥99%, ≥99% (GC) Formula Weight: 72.15g/mol Purity Grade: CHROMASOLV™ Explosion Limit: 0.083 Flash Point: −49°C (−56.2°F) Health Hazard 2: P210-P261-P273-P301 + P310-P331 Ignition Point: 500°F Linear Formula: CH3(CH2)3CH3 MDL Number: MFCD00009498 RTECS Number: RZ9450000 UN Number: UN1265 Vapor Density: 2.48 (vs air)
  spinner