Petroleum Ether
Ligroine, 100% Heptane (Laboratory), Fisher Chemical
Chemical Name or Material: Ligroine, Heptane 100% CAS: 142-82-5 Purity Grade: Laboratory Grade Molecular Formula: C7H16 Formula Weight: 100.2 MDL Number: MFCD00081849 Synonym: Petroleum Ether
Petroleum ether, ACS reagent, boiling range 40-60°C, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: ACS Reagent Grade MDL Number: MFCD00081849
Petroleum Ether (Pesticide), Fisher Chemical
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: Pesticide Grade Purity Grade Notes: Also meets ACS specifications MDL Number: MFCD00081849 UN Number: 1268 Synonym: Benzine
Petroleum Ether, Boiling Range 35-60°C, ACROS Organics™
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: Extra Pure MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum ether, extra pure, boiling range 100-140°C, ACROS Organics™
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 CAS: 111-65-9 CAS: 142-82-5 CAS: 108-87-2 CAS: 110-82-7 CAS: 73513-42-5 Purity Grade: Extra Pure MDL Number: MFCD00081849
Petroleum Ether, Extra Pure, Boiling range 40 to 60deg.C, Water <50ppm, AcroSeal™, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: Extra Dry MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum Ether, Optima™, Fisher Chemical
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: OPTIMA™ Purity Grade Notes: High purity extraction solvent for GC applications MDL Number: MFCD00081849 Synonym: Benzine
Petroleum ether, extra pure, boiling range 40-60°C, ACROS Organics™
Chemical Name or Material: Petroleum ether Name Note: Extra Dry CAS: 64742-49-0 Purity Grade: Extra Pure MDL Number: MFCD00081849
Petroleum ether, for analysis, boiling range 80-110°C, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: For analysis
Petroleum Ether Products for Science Education
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: ACS Reagent Grade Synonym: Benzine
Petroleum ether, for residue analysis, boiling range 40° to 60°C, ECD tested halocarbons free, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum ether Name Note: ECD Tested CAS: 64742-49-0 Purity Grade: For residue analysis MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum Ether, Liquid Products for Science Education
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: ACS Reagent Grade Physical Form: Liquid Synonym: Benzine
Petroleum ether, for residue analysis, 40° to 60°C,ECD tested lindane/DDT<5ng/L, res <0.0003%, ACROS Organics™
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: For residue analysis MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum Ether (Certified ACS), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: Certified ACS Reagent Grade MDL Number: MFCD00081849 Synonym: Benzine
Petroleum ether, for residue analysis, 40° to 60°C, for trace anal. of polyarom. hydrocarbons, ACROS Organics™
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: For residue analysis MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum ether 35/60, ACS grade, Alfa Aesar™
Chemical Name or Material: Petroleum ether 35/60 CAS: 8032-32-4 Purity Grade: ACS Grade Physical Form: Liquid MDL Number: MFCD00081849 Synonym: Ligroin
Ligroine, Hexane (Laboratory), Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Ligroine, Hexane (Laboratory) CAS: 110-54-3 Purity Grade: Laboratory Grade Molecular Formula: C6H14 Formula Weight: 86.18 MDL Number: MFCD00081849 Synonym: Hexanes
Petroleum ether 35/60, Alfa Aesar™
Chemical Name or Material: Petroleum ether 35/60 Name Note: HPLC Grade CAS: 8032-32-4 Physical Form: Liquid MDL Number: MFCD00081849
Petroleum ether 35/60, Environmental Grade, Alfa Aesar™
Chemical Name or Material: Petroleum ether 35/60 Name Note: Environmental Grade CAS: 8032-32-4 Purity Grade: Environmental Grade Physical Form: Liquid Assay Percent Range: >98% MDL Number: MFCD00081849
Petroleum Ether, Reagent ACS, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum Ether CAS: 64742-49-0 Purity Grade: ACS Reagent Grade MDL Number: MFCD00081849 Synonym: Benzine
Petroleum ether, for analysis, boiling range 60-95°C, ACROS Organics™
Chemical Name or Material: Petroleum ether Name Note: Boiling range 60° to 95°C CAS: 64742-49-0 Purity Grade: For analysis MDL Number: MFCD00081849 Synonym: Ligroine
Petroleum ether, extra pure, boiling range 60-95°C, ACROS Organics™ Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum ether CAS: 64742-49-0 Purity Grade: Extra Pure
Petroleum Ether UltimAR, Macron Fine Chemicals™
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4
Petroleum Ether ACS AR, Macron Fine Chemicals™
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Purity Grade: ACS Reagent Grade
Petroleum Ether, Macron Fine Chemicals™
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4 Synonym: Ligroin
Petroleum Ether AR, Macron Fine Chemicals™
Chemical Name or Material: Petroleum Ether CAS: 8032-32-4
Petroleum ether, Ultra Resi-analyzed, For ACS analysis, J.T.Baker™
Chemical Name or Material: Petroleum ether Name Note: For ACS analysis CAS: 64742-82-1 Physical Form: Liquid Molecular Formula: C6H14 Formula Weight: 86.18 Synonym: Benzin
Petroleum ether, Honeywell Riedel-de Haën™
CAS: 101316-46-5 Chemical Name or Material: Petroleum ether Purity Grade: CHROMASOLV™ Flash Point: −40°C (−40°F) Health Hazard 2: P210-P260-P280-P301 + P310-P370 + P378-P403 + P235 MDL Number: MFCD00081849 UN Number: UN1268
Petroleum Ether Non UV, EMD Millipore
Chemical Name or Material: Petroleum Ether Name Note: Non UV, Mixture CAS: 79-29-8 Physical Form: Liquid Linear Formula: Mixture Synonym: Ligroine
Petroleum Ether, ACS, EMD Millipore Available on GSA/VA Contract for Federal Government customers only.
Chemical Name or Material: Petroleum Ether Name Note: For ACS analysis CAS: 8032-32-4 Physical Form: Liquid Synonym: Ligroine
  spinner