AA ICP AES Standards
Aluminum Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.982 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
VeriSpec™ Cesium Standard for ICP, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical
CAS: 7647-17-8 Molecular Formula: ClCs Molecular Weight (g/mol): 168.355 InChI Key: AIYUHDOJVYHVIT-UHFFFAOYSA-M PubChem CID: 24293 ChEBI: CHEBI:63039 IUPAC Name: cesium;chloride SMILES: [Cl-].[Cs+]
Cesium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7789-18-6 Molecular Formula: CsNO3 Molecular Weight (g/mol): 194.909 InChI Key: NLSCHDZTHVNDCP-UHFFFAOYSA-N PubChem CID: 62674 IUPAC Name: cesium;nitrate SMILES: [N+](=O)([O-])[O-].[Cs+]
Copper Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-50-8 Molecular Formula: Cu Molecular Weight (g/mol): 63.546 InChI Key: RYGMFSIKBFXOCR-UHFFFAOYSA-N PubChem CID: 23978 ChEBI: CHEBI:30052 IUPAC Name: copper SMILES: [Cu]
Iron Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7439-89-6 Molecular Formula: Fe Molecular Weight (g/mol): 55.845 InChI Key: XEEYBQQBJWHFJM-UHFFFAOYSA-N PubChem CID: 23925 ChEBI: CHEBI:82664 IUPAC Name: iron SMILES: [Fe]
Yttrium Standard For ICP, 2% HNO3 in 1000μg/mL, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-65-5 Molecular Formula: Y Molecular Weight (g/mol): 88.906 InChI Key: VWQVUPCCIRVNHF-UHFFFAOYSA-N PubChem CID: 23993 ChEBI: CHEBI:33331 IUPAC Name: yttrium SMILES: [Y]
Magnesium Standard For ICP, 10,000μg/mL in 5% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 13446-18-9 Molecular Formula: H12MgN2O12 Molecular Weight (g/mol): 256.403 InChI Key: MFUVDXOKPBAHMC-UHFFFAOYSA-N PubChem CID: 202877 IUPAC Name: magnesium;dinitrate;hexahydrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Mg+2]
Magnesium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7439-95-4 Molecular Formula: Mg Molecular Weight (g/mol): 24.305 InChI Key: FYYHWMGAXLPEAU-UHFFFAOYSA-N PubChem CID: 5462224 ChEBI: CHEBI:25107 IUPAC Name: magnesium SMILES: [Mg]
Sulfur Standard For ICP, 10,000μg/mL in Water, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7783-20-2 Molecular Formula: H8N2O4S Molecular Weight (g/mol): 132.134 InChI Key: BFNBIHQBYMNNAN-UHFFFAOYSA-N PubChem CID: 6097028 ChEBI: CHEBI:62946 IUPAC Name: diazanium;sulfate SMILES: [NH4+].[NH4+].[O-]S(=O)(=O)[O-]
Boron AA Standard, 1mL = 1mg B (1,000ppm B)H3BO3 in H2O, Ricca Chemical
CAS: 10043-35-3 Molecular Formula: BH3O3 Molecular Weight (g/mol): 61.831 InChI Key: KGBXLFKZBHKPEV-UHFFFAOYSA-N PubChem CID: 7628 ChEBI: CHEBI:33118 IUPAC Name: boric acid SMILES: B(O)(O)O
Potassium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7447-40-7 Molecular Formula: ClK Molecular Weight (g/mol): 74.548 InChI Key: WCUXLLCKKVVCTQ-UHFFFAOYSA-M PubChem CID: 4873 ChEBI: CHEBI:32588 IUPAC Name: potassium;chloride SMILES: [Cl-].[K+]
Calcium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.078 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Platinum ICP Standard, 1mL = 10mg Pt (10,000ppm Pt), Ricca Chemical
CAS: 7440-06-4 Molecular Formula: Pt Molecular Weight (g/mol): 195.08 InChI Key: BASFCYQUMIYNBI-UHFFFAOYSA-N PubChem CID: 23939 ChEBI: CHEBI:33400 IUPAC Name: platinum SMILES: [Pt]
Sodium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Potassium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7447-40-7 Molecular Formula: ClK Molecular Weight (g/mol): 74.548 InChI Key: WCUXLLCKKVVCTQ-UHFFFAOYSA-M PubChem CID: 4873 ChEBI: CHEBI:32588 IUPAC Name: potassium;chloride SMILES: [Cl-].[K+]
Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 16919-19-0 Molecular Formula: F6H8N2Si Molecular Weight (g/mol): 178.153 InChI Key: ITHIMUMYFVCXSL-UHFFFAOYSA-P PubChem CID: 28145 IUPAC Name: diazanium;hexafluorosilicon(2-) SMILES: [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F
Sodium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-23-5 Molecular Formula: Na Molecular Weight (g/mol): 22.99 InChI Key: KEAYESYHFKHZAL-UHFFFAOYSA-N PubChem CID: 5360545 ChEBI: CHEBI:26708 IUPAC Name: sodium SMILES: [Na]
Cobalt Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.933 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Arsenic Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-38-2 Molecular Formula: As Molecular Weight (g/mol): 74.922 InChI Key: RQNWIZPPADIBDY-UHFFFAOYSA-N PubChem CID: 5359596 ChEBI: CHEBI:27563 IUPAC Name: arsenic SMILES: [As]
Boron Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Calcium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-70-2 Molecular Formula: Ca Molecular Weight (g/mol): 40.078 InChI Key: OYPRJOBELJOOCE-UHFFFAOYSA-N PubChem CID: 5460341 ChEBI: CHEBI:29320 IUPAC Name: calcium SMILES: [Ca]
Manganese Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7439-96-5 Molecular Formula: Mn Molecular Weight (g/mol): 54.938 InChI Key: PWHULOQIROXLJO-UHFFFAOYSA-N PubChem CID: 23930 ChEBI: CHEBI:35154 IUPAC Name: manganese SMILES: [Mn]
Strontium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-24-6 Molecular Formula: Sr Molecular Weight (g/mol): 87.62 InChI Key: CIOAGBVUUVVLOB-UHFFFAOYSA-N PubChem CID: 5359327 ChEBI: CHEBI:33324 IUPAC Name: strontium SMILES: [Sr]
Nickel Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-02-0 Molecular Formula: Ni Molecular Weight (g/mol): 58.693 InChI Key: PXHVJJICTQNCMI-UHFFFAOYSA-N PubChem CID: 935 ChEBI: CHEBI:28112 IUPAC Name: nickel SMILES: [Ni]
Tin Standard For ICP, 10,000μg/mL, SPEX CertiPrep™
CAS: 7440-31-5 Molecular Formula: Sn Molecular Weight (g/mol): 118.71 InChI Key: ATJFFYVFTNAWJD-UHFFFAOYSA-N PubChem CID: 5352426 ChEBI: CHEBI:32990 IUPAC Name: tin SMILES: [Sn]
Phosphorus Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7722-76-1 Molecular Formula: H6NO4P Molecular Weight (g/mol): 115.025 InChI Key: LFVGISIMTYGQHF-UHFFFAOYSA-N PubChem CID: 24402 ChEBI: CHEBI:62982 IUPAC Name: azanium;dihydrogen phosphate SMILES: [NH4+].OP(=O)(O)[O-]
Cadmium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7440-43-9 Molecular Formula: Cd Molecular Weight (g/mol): 112.414 InChI Key: BDOSMKKIYDKNTQ-UHFFFAOYSA-N PubChem CID: 23973 ChEBI: CHEBI:22977 IUPAC Name: cadmium SMILES: [Cd]
Mercury Standard For ICP, 1000μg/mL in 10% HNO3, Assurance Grade, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 7439-97-6 Molecular Formula: Hg Molecular Weight (g/mol): 200.592 InChI Key: QSHDDOUJBYECFT-UHFFFAOYSA-N PubChem CID: 23931 ChEBI: CHEBI:16170 IUPAC Name: mercury SMILES: [Hg]
  spinner