COD Standards
Chemical Oxygen Demand Standard, 300 ppm COD, Ricca Chemical
Chemical Name or Material: COD STANDARD
COD Standard, 1000ppm (1mL = 1mg COD) (Potassium Acid Phthalate in Water), Certified, 0.300mg/mL ±0.003, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735
COD Standard, 300ppm (1mL = 0.3mg COD) (Potassium Acid Phthalate in Water), Certified, 0.300mg/mL ±0.003, LabChem™ Available on GSA/VA Contract for Federal Government customers only.
CAS: 877-24-7 Molecular Formula: C8H5KO4 Molecular Weight (g/mol): 204.222 InChI Key: IWZKICVEHNUQTL-UHFFFAOYSA-M PubChem CID: 23676735
Sulfuric Acid-Silver Sulfate Reagent, for Chemical Oxygen Demand (COD) Analysis, Ricca Chemical
CAS: 7664-93-9 Molecular Formula: H2O4S Molecular Weight (g/mol): 98.072 InChI Key: QAOWNCQODCNURD-UHFFFAOYSA-N PubChem CID: 1118 ChEBI: CHEBI:26836
  spinner