Autosampler Vial Closure Accessories
DWK Life Sciences MicroLiter EZ-Decappers™ Hand Operated Vial Cappers and Decappers
MicroLiter EZ-Decappers™ Hand Operated Cappers and Decappers by MicroLiter are easy-to-use tools for crimping and decrimping chromatography vial closures.
Fisherbrand™ 150μL Clear Polyspring Insert
Fisherbrand™ 150μL Clear Polyspring Inserts features superior glass design and a write-on patch with graduation for convenient sample identification.
Fisherbrand™ 400μL Clear Microsert Insert
Unique features for wide opening screw vials such as convenient sample identification, wide neck opening design that allows for easy filling, and superior glass.
Fisherbrand™ 200μL Flat Bottom Microsert Insert
Fisherbrand™ 200μL Flat Bottom Microsert Insert features superior glass design and a write-on patch with graduation for convenient sample identification.
Simport™ Scientific Capinsert™ For Cryovial Tubes
Inserts fit precisely into the cap of the Cryovial™ for color identification
Fisherbrand™ 300μL Polypropolene Insert
Unique features for wide opening screw vials such as convenient sample identification, wide neck opening design that allows for easy filling,and superior glass.
Restek™ Dual Vespel™ Ring Inlet Seals
Extend column lifetime by preventing oxygen from reaching the column with these inlet seals. Restek™ Dual Vespel™ Ring Inlet Seals eliminate the need for a washer and ensure very little torque is needed to make a leak-tight seal.
Fisherbrand™ 350μL Clear Insert
Includes many unqiue features for thread vials such as convenient sample identification, wide neck opening design for easy filling, and polyspring inserts.
  spinner