Alkylation Reagent
Thermo Scientific™ MethElute™ Reagent (TMPAH) Available on GSA/VA Contract for Federal Government customers only.
For accurate, sensitive on-column methylation use Thermo Scientific™ MethElute™ Reagent (TMPAH).
Thermo Scientific™ Methylate Reagent (DMFDMA)
For easy, effective preparation of methyl esters from fatty acids and amino acids use Thermo Scientific™ Methylate Reagent (DMFDMA).
Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Derivatize carboxyl acids, phenols, and sulfonamides for electron-capture detection (ECD) GC analysis with the Thermo Scientific™ Pentafluorobenzyl Bromide (PFBBr) Alkylation Reagent.
  spinner