Silylation Reagents
Thermo Scientific™ BSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1% TMCS Silylation Reagent is derivatizing compounds.
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.
Thermo Scientific™ BSTFA Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Obtain excellent GC separations with Thermo Scientific™ BSTFA Silylation Reagent.
Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent Available on GSA/VA Contract for Federal Government customers only.
Employ a catalyzed silylation reagent formulation, for derivatization of hydroxyls with the Thermo Scientific™ Tri-Sil TBT (TMSI:BSA:TMCS) Reagent.
Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent Available on GSA/VA Contract for Federal Government customers only.
Employ a reagent-catalyst solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil HTP (HDMS:TMCS:Pyridine) Reagent,
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ Methoxamine (MOX) Reagent Available on GSA/VA Contract for Federal Government customers only.
Prepare oximes of steroids and ketoacids prior to silylation with the Thermo Scientific™ Methoxamine (MOX) Reagent.
Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent Available on GSA/VA Contract for Federal Government customers only.
Derivatize hydroxyls compounds, particularly carbohydrates with Thermo Scientific™ Tri-Sil TP (TMSI:Pyridine) Reagent.
Thermo Scientific™ TMSI Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Employ the strongest silylator available for carbohydrates and steroids with the Thermo Scientific™ TMSI Silylation Reagent.
Thermo Scientific™ HMDS (Hexamethyldisilazane) Available on GSA/VA Contract for Federal Government customers only.
Greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent Available on GSA/VA Contract for Federal Government customers only.
Employ a reagent-solvent mixture, for one-step derivatization with the Thermo Scientific™ Tri-Sil BP (BSA:Pyridine) Reagent,
Thermo Scientific™ BSA Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Produce volatile, stable TMS derivatives quickly and quantitatively under mild conditions with Thermo Scientific™ BSA Silylation Reagent.
Thermo Scientific™ TMCS Silylation Reagent Available on GSA/VA Contract for Federal Government customers only.
Catalyze the derivatization of difficult-to-silylate compounds using Thermo Scientific™ TMCS Silylation Reagent.
  spinner