Gas Chromatography Port Injection Liners
Thermo Scientific™ LinerGOLD™ GC Liners Available on GSA/VA Contract for Federal Government customers only.
Use the gold-standard LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Thermo Scientific™ Injection Port Liners for Agilent instruments Available on GSA/VA Contract for Federal Government customers only.
Use these injection port liners to optimize performance of Agilent GC instruments.
Straight Inlet Liners with Wool, I.D.4mm from Restek™
For use with Agilent GCs equipped with split or splitless inlets.
SGE™ GC Inlet FocusLiners™
Results in lower mass discrimination
Inlet Liners for Splitless Injection—Gooseneck Splitless with FS Wool from Restek™
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems.
Restek™ Inlet Liners for Split Injection - Split Precision™ Liner
For Agilent GCs equipped with split/splitless inlets.
Restek™ Inlet Liners for Splitless Injection—Gooseneck Splitless
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Restek™ Inlet Liners for Splitless Injection - Recessed Gooseneck with FS Wool
For Agilent GCs equipped with split/splitless inlets.
Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Split Liner
Specifically designed to ensure a highly inert GC pathway for basic compounds
Restek™ CarboFrit-Packed Inlet Liners for Agilent GCs
For Agilent GCs equipped with split/splitless inlets.
Restek™ Inlet Liners for Split Injection - Siltek™ Split Precision Liner
For Agilent GCs equipped with split/splitless inlets.
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments Available on GSA/VA Contract for Federal Government customers only.
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Restek™ Split Liners for Shimadzu GCs
For Shimadzu GCs equipped with split/splitless inlets.
Restek™ Straight Inlet Liners for Splitless Injection
Used for samples containing a narrow molecular weight distribution and for those not prone to thermal decomposition
Trajan™ Agilent Split / Splitless Tapered FocusLiner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Restek™ Inlet Liners for Splitless Injection - Splitless (Quartz)
Useful for splitless mode of operation and slow injections, small (<2μL) sample volumes, and high-boiling point solvent systems
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments Available on GSA/VA Contract for Federal Government customers only.
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Restek™ Inlet Liners for Split Injection
Fully deactivated to prevent degradation or adsorption of active compounds
SGE™ GC Inlet Tapered FocusLiners™ Split/Splitless
Results in lower mass discrimination
Restek™ Inlet Liners for Splitless Injection - Double Gooseneck Splitless
For Agilent GCs equipped with split/splitless inlets.
Restek™ Splitless Liners for PerkinElmer GCs: Auto SYS Splitless
For PerkinElmer Auto SYS and Clarus GCs equipped with split/splitless inlets.
Restek™ Inlet Liners for Basic Organic Analysis for Agilent™ GCs: Gooseneck Splitless
Specifically designed to ensure a highly inert GC pathway for basic compounds
SGE™ Split Liner With Quartz Wool
For various GC Instruments
Restek™ Split/Splitless Liners for Shimadzu: 17A Split/Splitless w/Wool
For Shimadzu 17A, 2010, and 2014 GCs equipped with split/splitless inlets.
Restek™ CIS4 and PTV Liners for Agilent: Baffled Glass Inlet Liner
For Agilent GCs equipped with CIS4/PTV inlets.
Restek™ Split Liners for TRACE PTV
For Thermo TRACE GCs equipped with PTV inlets.
  spinner