Chromatography Ferrules
Thermo Scientific™ SilTite™ Ferrules Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ SilTite Ferrules are for use with capillary column connectors.
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments Available on GSA/VA Contract for Federal Government customers only.
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ Ferrules.
Thermo Scientific™ SLIPFREE Ferrules Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Thermo Scientific™ Finger Tite Ferrules Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific™ Ferrules for Agilent™ Instruments
Ensure reliable connections for Agilent™ GC instruments with these Thermo Scientific™ Ferrules for Agilent Instruments.
Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments Available on GSA/VA Contract for Federal Government customers only.
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments.
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules Available on GSA/VA Contract for Federal Government customers only.
Mechanically robust for a long lifetime and are compatible with GC/MS
Thermo Scientific™ Brass Nuts for 15% Graphite/85% Vespel Ferrules
Thermo Scientific™ Brass Nuts for 15% Graphite/85% Vespel Ferrules are mechanically robust for a long lifetime and are compatible with GC/MS.
Thermo Scientific™ 100% Graphite Ferrules Available on GSA/VA Contract for Federal Government customers only.
Composed of a soft material that is porous to oxygen, making them suitable for most applications except GC/MS interface connections
Restek™ Reducing Ferrules: Graphite
Made from high-purity, high-density graphite.
Restek™ Vu-Union Vespel/Graphite Ferrules
Use with Vacuum Vu-Union connectors.
SGE™ 100% Graphite Ferrules
Use in Agilent Technologies™ (HP) detectors and injectors at atmospheric pressure (e.g., FID)
SGE™ 15% Graphite/85% Vespel™ Reducing Ferrules
Mechanically robust for long lifetimes
SGE™ 15% Graphite/85% Vespel™ Straight Ferrules
Mechanically robust for long lifetimes
Restek™ Compact Ferrules: Graphite
For Agilent™ 5890, 6890, and 6850 columns
Restek™ Compact Ferrules: Vespel/Graphite
For Agilent™ 5890, 6890, and 6850 columns
Restek™ PTFE Fluorocarbon Resin Ferrules
One-piece design requires no back ferrule.
Restek™ One-Piece Fused Silica Adaptor Ferrule
Fused silica adaptor ferrules made from Valcon polyimide or virgin polyimide.
Restek™ Ferrules for Capillary Columns: Vespel/Graphite
Made of high-purity materials that will not contribute background impurities for sensitive and selective detectors
Restek™ Encapsulated Ferrules
For 0.0625 in. compression fittings.
SGE™ 15% Graphite/85% Vespel™ Ferrules
Mechanically robust for long lifetimes
SGE™ 15% Graphite/85% Vespel™ Two-Hole Ferrules
Mechanically robust for long lifetimes
  spinner