Chromatography Septa
Thermo Scientific™ Septa Available on GSA/VA Contract for Federal Government customers only.
A wide range to cover most applications
Thermo Scientific™ Septa Available on GSA/VA Contract for Federal Government customers only.
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Restek™ BTO™ Septa: With CenterGuide
Plasma coating eliminates sticking in the injection port
Trajan™ EC GC Septa
EC GC septa is designed for long lifetime and low injection port adhesion
Restek™ BTO™ Septa: Shimadzu Plug
Plasma coating eliminates sticking in the injection port
Hamilton™ GC Septa
Three Layer Septa for operating temperatures up to 200° C
Thermo Scientific™ Septum Holder Kit Non-distribution product as customer accommodation.
Thermo Scientific™ Septum Holder Kit is for use with the Thermo Scientific™ Trace 2000 Series Gas Chromatograph.
Trajan™ HT GC Septa
HT GC septa is a bleed and temperature optimized material
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Trajan Scientific and Medical™ GC Septum Ht Type 9A
Retains softness and pierceability at high temperatures and low injection port adhesion.
SGE™ MN Silicone Septa
Use these septa for a variety of applications. SGE Analytical Science™ MN Silicone Septa are made with a high grade silicone material to minimize needle coring.
  spinner