Packings for Chromatography Columns
GE Healthcare Glutathione Sepharose™ 4B Media Available on GSA/VA Contract for Federal Government customers only.
Use for high capacity purification of Glutathione S-Transferase (GST) tagged fusion proteins. GE Healthcare Life Sciences™ Glutathione Sepharose™ 4B Media requires only a single-step for purification.
Applied Biosystems™ 4-Capillary Array For 3130/3100-Avant™ Genetic Analyzer
Use for fragment analysis and sequencing. Usage per array is 100 runs.
GE Healthcare Benzamidine Sepharose™ 4 Fast Flow, High Sub Media Available on GSA/VA Contract for Federal Government customers only.
For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases and thrombin
GE Healthcare HiTrap™ Capto™ IEX Selection Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for use with syringe, peristalic pump, or chromatography systems such as AKTA design for convenient media screening and optimization studies
Selecto™ Preparative and Classic Column Chromatography Active Silica
60Å gels made from identical starting material, making scale-up easier
Selecto™ Preparative and Classic Column Chromatography Silica
60Å gels made from identical starting material, making scale-up easier
  spinner