SPE Columns, Discs, and Cartridges
Thermo Scientific™ SOLA™ SPE Cartridges Available on GSA/VA Contract for Federal Government customers only.
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Thermo Scientific™ HyperSep™ Silica Cartridges Available on GSA/VA Contract for Federal Government customers only.
Retain analytes in nonpolar matrices, and separate compounds of very similar structure with these silica SPE cartridges.
Thermo Scientific™ SMART Digest™ Trypsin Kits
Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.
Thermo Scientific™ SMART Digest™ Chymotrypsin Kits
Obtain fast, simple, and highly reproducible chymotrypsin protein digestion for peptide characterization and quantitation in biopharmaceutical research.
Thermo Scientific™ SOLAµ™ SPE Plates
Remove variability and achieve highly consistent results. High reproducibility of Thermo Scientific™ SOLAµ™ SPE plates deliver greater sample success at low elution volumes.
Thermo Scientific™ HyperSep™ Florisil Cartridges Available on GSA/VA Contract for Federal Government customers only.
Isolate polar compounds from nonpolar matrices with these Florisil SPE cartridges, which are especially well-suited for extraction of chlorinated pesticides.
Thermo Scientific™ HyperSep™ C18 Cartridges Available on GSA/VA Contract for Federal Government customers only.
Get improved retention for most nonpolar compounds and organic analytes in aqueous matrices with these hydrophobic reversed phase C18 cartridges.
Avantor™ BAKERBOND Speedisk™ Octadecyl (C18) Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Octadecyl (C18) Extraction Disks are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Avantor™ BAKERBOND™ spe Octadecyl (C18) Disposable Extraction Columns
Define and control critical surface chemistry parameters to ensure performance consistency. Avantor BAKERBOND spe Octadecyl (C18) Disposable Extraction Columns, are designed with standard geometries to adapt to most popular automated liquid handling systems for extracting non-ionic, non-polar to moderately polar analytes.
Thermo Scientific™ HyperSep™ Retain PEP Cartridges Available on GSA/VA Contract for Federal Government customers only.
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP Cartridges.
Thermo Scientific™ HyperSep™ Hypercarb™ SPE Cartridges Available on GSA/VA Contract for Federal Government customers only.
Unique graphite carbon-based SPE cartridges offer retention and separation of polar compounds where silica and resin stationary phases fail.
Thermo Scientific™ HyperSep™ Diol Cartridges Available on GSA/VA Contract for Federal Government customers only.
Obtain excellent normal phase extraction of polar compounds with these diol SPE cartridges.
Thermo Scientific™ HyperSep™ SCX Cartridges Available on GSA/VA Contract for Federal Government customers only.
Obtain excellent performance for extraction of charged basic compounds with these strong cation exchanges SPE cartridges.
Thermo Scientific™ HyperSep™ SAX Cartridges Available on GSA/VA Contract for Federal Government customers only.
Obtain excellent performance for extraction of carboxylic and other weak acids with these strong anion exchanger SPE cartridges.
Restek™ Resprep™-C18 and Resprep™-C8 SPE Disks
Extract analytes using high flow rates with significantly less clogging
Restek™ Resprep Empty SPE Tubes for Packing
For use with Resprep™ SPE system.
Avantor™ BAKERBOND Speedisk™ Oil and Grease Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Oil and Grease Extraction Disk are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Restek™ Resprep Empty SPE Frits for Packing
For use with Resprep™ SPE Tubes.
Restek™ Filters for ASE 200 Extraction Cells
Fit 11 mL, 22 mL, and 33 mL cells.
Thermo Scientific™ HyperSep™ C8 Cartridges Available on GSA/VA Contract for Federal Government customers only.
Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 cartridges offer excellent performance.
Avantor™ BAKERBOND Speedisk™ H2O-Philic DVB Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disk H2O-Philic DVB are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Restek™ Resprep SPE Connectors and End Cap
For use with Resprep™ SPE Tubes.
Restek™ Resprep Empty SPE Caps for Packing
For use with Resprep™ SPE Tubes.
Thermo Scientific™ HyperSep™ Aminopropyl Cartridges Available on GSA/VA Contract for Federal Government customers only.
Obtain excellent retention of drugs, metabolites and structural isomers with these aminopropyl SPE cartridges that allow both polar and anion exchange interactions.
Restek™ CarboPrep Cartridges
Our manufacturing process minimizes variability and improves recovery and cleanup procedures.
Restek™ Normal Phase Resprep SPE Cartridges
Hydrophilic (polar) adsorbents used to extract hydrophilic analytes from nonpolar matrices.
Avantor™ BAKERBOND Speedisk™ H2O-Phobic DVB Extraction Disk
Maximize capacity, adsorption and SPE speed. Avantor BAKERBOND Speedisk Extraction Disk H2O-Phobic DVB are neither cartridge nor membrane that provides an excellent SPE tool for environmental samples and combines a thin bed of BAKERBOND sorbent with a special laminar configuration for testing drinking water, ground water, waste water, solid waste or process streams.
Restek™ Bonded Reversed Phase Resprep SPE Cartridges
Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.
  spinner